Assay Details
Gene Symbol: POU6F2-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK023033.1 | - | 3 - 4 | 402 | 189 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
VPS41, HOPS complex subunit |
VPS41 |
HVPS41, HVSP41, hVps41p |
-79.4kb | Sense | Chr. 7 - 38722974 - 38909200 on NCBI Build GRCh38 |
Downstream | POU6F2 antisense RNA 1 | POU6F2-AS1 | 402.4kb | Sense | Chr. 7 - 39404598 - 39406346 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() |
