Assay Details
Gene Symbol: SEPT14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207366.2 | NP_997249.2 | 1 - 2 | 104 | 60 | |
GenBank mRNA | AK301928.1 | - | 1 - 2 | 67 | 60 | |
DC400351.1 | - | 1 - 2 | 104 | 60 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC07001405.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK301928.1,NM_207366.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR07021803.hg.1 | TC07001405.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07021803.hg.1 | ENST00000388975.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07021803.hg.1 | ENST00000477628.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07021803.hg.1 | NM_207366.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011156.hg.1 | ENST00000388975.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011156.hg.1 | ENST00000388975.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011156.hg.1 | ENST00000388975.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011156.hg.1 | ENST00000388975.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011156.hg.1 | ENST00000388975.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011156.hg.1 | ENST00000388975.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011156.hg.1 | ENST00000388975.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011156.hg.1 | ENST00000388975.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011156.hg.1 | ENST00000388975.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011156.hg.1 | ENST00000388975.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011156.hg.1 | ENST00000388975.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011156.hg.1 | ENST00000388975.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011156.hg.1 | ENST00000388975.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011156.hg.1 | ENST00000477628.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011156.hg.1 | ENST00000477628.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011156.hg.1 | ENST00000477628.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011156.hg.1 | ENST00000477628.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011156.hg.1 | ENST00000477628.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011156.hg.1 | NM_207366.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011156.hg.1 | NM_207366.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011156.hg.1 | NM_207366.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011156.hg.1 | NM_207366.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011156.hg.1 | NM_207366.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011156.hg.1 | NM_207366.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011156.hg.1 | NM_207366.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011156.hg.1 | NM_207366.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011156.hg.1 | NM_207366.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011156.hg.1 | NM_207366.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011156.hg.1 | NM_207366.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011156.hg.1 | NM_207366.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011156.hg.1 | NM_207366.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07021801.hg.1 | TC07001405.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07021801.hg.1 | ENST00000388975.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07021801.hg.1 | ENST00000477628.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07021801.hg.1 | NM_207366.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07021804.hg.1 | TC07001405.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07021804.hg.1 | ENST00000388975.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07021804.hg.1 | ENST00000477628.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07021804.hg.1 | NM_207366.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07021805.hg.1 | TC07001405.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07021805.hg.1 | ENST00000388975.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07021805.hg.1 | NM_207366.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700179727.hg.1 | ENST00000388975.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700179727.hg.1 | ENST00000477628.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700179727.hg.1 | NM_207366.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700179727.hg.1 | ENST00000388975.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700179727.hg.1 | ENST00000477628.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700179727.hg.1 | NM_207366.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700179727.hg.1 | TC0700011156.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700179730.hg.1 | ENST00000388975.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700179730.hg.1 | NM_207366.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700179730.hg.1 | ENST00000388975.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700179730.hg.1 | NM_207366.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700179730.hg.1 | TC0700011156.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700179729.hg.1 | ENST00000388975.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700179729.hg.1 | ENST00000477628.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700179729.hg.1 | NM_207366.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700179729.hg.1 | ENST00000388975.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700179729.hg.1 | ENST00000477628.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700179729.hg.1 | NM_207366.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700179729.hg.1 | TC0700011156.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700179725.hg.1 | ENST00000388975.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700179725.hg.1 | ENST00000477628.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700179725.hg.1 | NM_207366.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700179725.hg.1 | ENST00000388975.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700179725.hg.1 | ENST00000477628.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700179725.hg.1 | NM_207366.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700179725.hg.1 | TC0700011156.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0700011156.hg.1 | ENST00000388975.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011156.hg.1 | ENST00000388975.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011156.hg.1 | ENST00000388975.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011156.hg.1 | ENST00000388975.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011156.hg.1 | ENST00000388975.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011156.hg.1 | ENST00000388975.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011156.hg.1 | ENST00000388975.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011156.hg.1 | ENST00000388975.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011156.hg.1 | ENST00000388975.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011156.hg.1 | ENST00000388975.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011156.hg.1 | ENST00000388975.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011156.hg.1 | ENST00000388975.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011156.hg.1 | ENST00000388975.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011156.hg.1 | ENST00000477628.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011156.hg.1 | ENST00000477628.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011156.hg.1 | ENST00000477628.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011156.hg.1 | ENST00000477628.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011156.hg.1 | ENST00000477628.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011156.hg.1 | NM_207366.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011156.hg.1 | NM_207366.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011156.hg.1 | NM_207366.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011156.hg.1 | NM_207366.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011156.hg.1 | NM_207366.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011156.hg.1 | NM_207366.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011156.hg.1 | NM_207366.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011156.hg.1 | NM_207366.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011156.hg.1 | NM_207366.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011156.hg.1 | NM_207366.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011156.hg.1 | NM_207366.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011156.hg.1 | NM_207366.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011156.hg.1 | NM_207366.2 | Refseq |
