Assay Details
| Gene Symbol: ANO5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142649.1 | NP_001136121.1 | 15 - 16 | 1943 | 155 | |
| NM_213599.2 | NP_998764.1 | 15 - 16 | 1946 | 155 | ||
| XM_005252820.3 | XP_005252877.2 | 14 - 15 | 2050 | 155 | ||
| XM_005252821.3 | XP_005252878.2 | 14 - 15 | 2047 | 155 | ||
| XM_005252822.4 | XP_005252879.1 | 15 - 16 | 1831 | 155 | ||
| XM_011519949.2 | XP_011518251.1 | 13 - 14 | 1999 | 155 | ||
| GenBank mRNA | AB125267.1 | - | 15 - 16 | 1946 | 155 | |
| AL832374.1 | - | 15 - 16 | 1968 | 155 | ||
| AL833271.1 | - | 15 - 16 | 1967 | 155 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000261.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100010338.hg.1 | ENST00000526935.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010338.hg.1 | ENST00000526935.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000532043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000532043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000532043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000532043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000532043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003519.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003519.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003519.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003519.hg.1 | TC11000261.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1100010338.hg.1 | ENST00000526935.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010338.hg.1 | ENST00000526935.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100149848.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100149848.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100149848.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100149848.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100149848.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100149848.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100149848.hg.1 | TC1100007063.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000532043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000532043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000532043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000532043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000532043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |

ANO5
Genomic Map