Assay Details
| Gene Symbol: MIR4500HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033829.1 | 1 - 2 | 122 | 133 | ||
| GenBank mRNA | AF339814.1 | - | 1 - 2 | 136 | 133 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13000771.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR13008814.hg.1 | ENST00000436290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13008814.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13008814.hg.1 | TC13000771.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR13008818.hg.1 | ENST00000436290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13008818.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13008818.hg.1 | TC13000771.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000453832.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000453832.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000453832.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000453832.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000453832.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000453832.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000606590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000606590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000606590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000606590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000607309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000607309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000607309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000607309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300163055.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163055.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300163055.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163055.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300163055.hg.1 | TC1300009405.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000453832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000453832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000453832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000453832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000453832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000453832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000606590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000606590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000606590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000606590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000607309.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000607309.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000607309.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000607309.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300163063.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163063.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163063.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163063.hg.1 | ENST00000453832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163063.hg.1 | ENST00000606590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163063.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300163063.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163063.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163063.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163063.hg.1 | ENST00000453832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163063.hg.1 | ENST00000606590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163063.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300163063.hg.1 | TC1300009405.hg.1 | Clariom D TC |

MIR4500HG
Genomic Map