Assay Details
Gene Symbol: NIFK-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_037857.1 | 2 - 3 | 946 | 69 | ||
GenBank mRNA | AK098264.1 | - | 2 - 3 | 472 | 69 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02000768.hg.1
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02012027.hg.1 | TC02000768.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02012027.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02012027.hg.1 | NR_037857.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000413904.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000413904.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000413904.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000413904.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000413904.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000413904.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000413904.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000413904.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000413904.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000413904.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000413904.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000413904.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000414554.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000414554.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000414554.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000414554.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000414554.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000414554.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000414554.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000414554.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000414554.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000414554.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000414554.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000414554.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000414554.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000414554.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000414554.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000414554.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000414554.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000414554.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000439321.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000439321.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000439321.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000439321.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000439321.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000439321.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000439321.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000447668.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000447668.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000447668.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000447668.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000447668.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000447668.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | ENST00000447668.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | NR_037856.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | NR_037856.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | NR_037856.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | NR_037856.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | NR_037856.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | NR_037856.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | NR_037856.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | NR_037857.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | NR_037857.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | NR_037857.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | NR_037857.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | NR_037857.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | NR_037857.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | NR_037857.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | NR_037857.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | NR_037857.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | NR_037857.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | NR_037857.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | NR_037857.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | NR_037857.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | NR_037857.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | NR_037857.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | NR_037858.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | NR_037858.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | NR_037858.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | NR_037858.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | NR_037858.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | NR_037858.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | NR_037858.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | NR_037858.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | NR_037858.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | NR_037858.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | NR_037858.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009128.hg.1 | NR_037858.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000413904.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000413904.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000413904.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000413904.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000413904.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000413904.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000413904.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000413904.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000413904.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000413904.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000413904.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000413904.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000414554.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000414554.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000414554.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000414554.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000414554.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000414554.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000414554.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000414554.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000414554.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000414554.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000414554.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000414554.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000414554.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000414554.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000414554.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000414554.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000414554.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000414554.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000439321.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000439321.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000439321.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000439321.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000439321.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000439321.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000439321.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000447668.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000447668.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000447668.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000447668.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000447668.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000447668.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | ENST00000447668.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | NR_037856.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | NR_037856.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | NR_037856.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | NR_037856.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | NR_037856.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | NR_037856.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | NR_037856.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | NR_037857.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | NR_037857.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | NR_037857.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | NR_037857.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | NR_037857.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | NR_037857.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | NR_037857.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | NR_037857.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | NR_037857.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | NR_037857.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | NR_037857.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | NR_037857.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | NR_037857.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | NR_037857.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | NR_037857.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | NR_037858.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | NR_037858.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | NR_037858.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | NR_037858.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | NR_037858.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | NR_037858.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | NR_037858.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | NR_037858.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | NR_037858.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | NR_037858.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | NR_037858.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009128.hg.1 | NR_037858.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200164018.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200164018.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200164018.hg.1 | NR_037857.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200164018.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200164018.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200164018.hg.1 | NR_037857.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200164018.hg.1 | TC0200009128.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200164017.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200164017.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200164017.hg.1 | NR_037857.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200164017.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200164017.hg.1 | ENST00000419902.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200164017.hg.1 | NR_037857.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200164017.hg.1 | TC0200009128.hg.1 | Clariom D TC |
