Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001927.hg.1 |
ENST00000449427.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001927.hg.1 |
NR_038394.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001927.hg.1 |
TC20000148.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001926.hg.1 |
NR_038394.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001926.hg.1 |
TC20000148.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001928.hg.1 |
ENST00000449427.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001928.hg.1 |
NR_038394.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001928.hg.1 |
TC20000148.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006921.hg.1 |
ENST00000449427.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006921.hg.1 |
ENST00000449427.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006921.hg.1 |
ENST00000449427.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006921.hg.1 |
ENST00000449427.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006921.hg.1 |
ENST00000449427.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006921.hg.1 |
ENST00000449427.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006921.hg.1 |
ENST00000449427.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006921.hg.1 |
ENST00000449427.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006921.hg.1 |
ENST00000449427.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006921.hg.1 |
ENST00000449427.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006921.hg.1 |
NR_038394.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006921.hg.1 |
NR_038394.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006921.hg.1 |
NR_038394.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006921.hg.1 |
NR_038394.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006921.hg.1 |
NR_038394.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006921.hg.1 |
NR_038394.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000147386.hg.1 |
ENST00000449427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000147386.hg.1 |
ENST00000449427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000147386.hg.1 |
NR_038394.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000147386.hg.1 |
TC2000006921.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000147386.hg.1 |
ENST00000449427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000147386.hg.1 |
ENST00000449427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000147386.hg.1 |
NR_038394.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006921.hg.1 |
ENST00000449427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006921.hg.1 |
ENST00000449427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006921.hg.1 |
ENST00000449427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006921.hg.1 |
ENST00000449427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006921.hg.1 |
ENST00000449427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006921.hg.1 |
ENST00000449427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006921.hg.1 |
ENST00000449427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006921.hg.1 |
ENST00000449427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006921.hg.1 |
ENST00000449427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006921.hg.1 |
ENST00000449427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006921.hg.1 |
NR_038394.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006921.hg.1 |
NR_038394.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006921.hg.1 |
NR_038394.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006921.hg.1 |
NR_038394.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006921.hg.1 |
NR_038394.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006921.hg.1 |
NR_038394.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000147385.hg.1 |
ENST00000449427.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000147385.hg.1 |
NR_038394.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000147385.hg.1 |
TC2000006921.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000147385.hg.1 |
ENST00000449427.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000147385.hg.1 |
NR_038394.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000147388.hg.1 |
ENST00000449427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000147388.hg.1 |
ENST00000449427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000147388.hg.1 |
NR_038394.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000147388.hg.1 |
TC2000006921.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000147388.hg.1 |
ENST00000449427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000147388.hg.1 |
ENST00000449427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000147388.hg.1 |
NR_038394.1 |
Refseq |