Assay Details
Gene Symbol: LINC01432 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038394.1 | 2 - 3 | 443 | 123 | ||
GenBank mRNA | BC038192.1 | - | 2 - 3 | 443 | 123 | |
BE789145.1 | - | 2 - 3 | 422 | 123 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | paired box 1 | PAX1 |
HUP48, OFC2 |
-350.0kb | Sense | Chr. 20 - 21705659 - 21718486 on NCBI Build GRCh38 |
Downstream | long intergenic non-protein coding RNA 1427 | LINC01427 | 206.8kb | Anti-sense | Chr. 20 - 22280228 - 22282801 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC20000148.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11112993 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756994 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756907 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756968 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17179372 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756924 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756934 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756903 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113033 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113062 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756963 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756910 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31757006 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113114 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113044 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756886 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113125 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756943 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113009 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31757038 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11112996 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756985 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756898 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756911 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756913 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11112984 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756883 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756912 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756882 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113001 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113063 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756946 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113078 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31757025 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756920 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113106 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31757021 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113046 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31757034 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113135 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11112983 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17179358 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113004 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113092 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113017 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17179368 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756885 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756980 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113136 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756970 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31757018 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31757013 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113018 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31757015 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756918 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756895 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17179370 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31757014 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17179383 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756989 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113053 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113074 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113072 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17179371 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756919 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756999 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31757028 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756893 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113049 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756961 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756969 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756921 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113091 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11112982 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113008 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756965 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113042 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31757029 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31757017 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756973 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113034 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31757035 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756904 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31757022 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11112994 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756896 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113010 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756927 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113043 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756981 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113090 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113047 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31757000 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756931 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17179376 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756998 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113080 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756964 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113094 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113123 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113086 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756975 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113083 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113011 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756960 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756953 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756925 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31757003 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756990 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113025 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756888 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31757032 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756916 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113054 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31757023 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11112986 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113029 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113036 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11112988 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756935 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113024 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113041 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113100 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113107 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113134 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756936 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113119 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756945 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113109 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756997 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113085 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756899 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31756889 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11113103 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 51845 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 190575 | AK314848.1 | genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 190575 | AX721132.1 | genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 190575 | BC028974.1 | genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 190575 | BX537601.1 | genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 190575 | EF534355.1 | genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 190575 | NM_173480.1 | refseq_mrna_human_NM |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 422236 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 219514 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 770866 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 939156 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 890244 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 501236 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3875565 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 56331 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3875565 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3875565 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P21099872 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 155802 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P21099887 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P21099872 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P41292647 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P21099845 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P718918 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P21099872 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P21099845 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR20001930.hg.1 | ENST00000449427.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20001930.hg.1 | NR_038394.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20001930.hg.1 | TC20000148.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR20001931.hg.1 | NR_038394.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20001931.hg.1 | TC20000148.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC2000006921.hg.1 | ENST00000449427.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006921.hg.1 | ENST00000449427.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006921.hg.1 | ENST00000449427.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006921.hg.1 | ENST00000449427.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006921.hg.1 | ENST00000449427.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006921.hg.1 | ENST00000449427.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006921.hg.1 | ENST00000449427.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006921.hg.1 | ENST00000449427.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006921.hg.1 | ENST00000449427.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006921.hg.1 | ENST00000449427.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006921.hg.1 | NR_038394.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006921.hg.1 | NR_038394.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006921.hg.1 | NR_038394.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006921.hg.1 | NR_038394.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006921.hg.1 | NR_038394.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006921.hg.1 | NR_038394.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20001929.hg.1 | ENST00000449427.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20001929.hg.1 | NR_038394.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20001929.hg.1 | TC20000148.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR2000147392.hg.1 | ENST00000449427.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147392.hg.1 | ENST00000449427.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147392.hg.1 | NR_038394.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000147392.hg.1 | TC2000006921.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000147392.hg.1 | ENST00000449427.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147392.hg.1 | ENST00000449427.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147392.hg.1 | NR_038394.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000147393.hg.1 | ENST00000449427.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147393.hg.1 | NR_038394.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000147393.hg.1 | TC2000006921.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000147393.hg.1 | ENST00000449427.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147393.hg.1 | NR_038394.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006921.hg.1 | ENST00000449427.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006921.hg.1 | ENST00000449427.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006921.hg.1 | ENST00000449427.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006921.hg.1 | ENST00000449427.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006921.hg.1 | ENST00000449427.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006921.hg.1 | ENST00000449427.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006921.hg.1 | ENST00000449427.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006921.hg.1 | ENST00000449427.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006921.hg.1 | ENST00000449427.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006921.hg.1 | ENST00000449427.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006921.hg.1 | NR_038394.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006921.hg.1 | NR_038394.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006921.hg.1 | NR_038394.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006921.hg.1 | NR_038394.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006921.hg.1 | NR_038394.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006921.hg.1 | NR_038394.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000147390.hg.1 | ENST00000449427.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147390.hg.1 | ENST00000449427.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147390.hg.1 | NR_038394.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000147390.hg.1 | TC2000006921.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000147390.hg.1 | ENST00000449427.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147390.hg.1 | ENST00000449427.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147390.hg.1 | NR_038394.1 | Refseq |
Human Genome U133 Plus 2.0 Array | 1563246_at | - | - |
Human Genome U133 Plus 2.0 Array | 1563246_PM_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1563246_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1563246_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1563246_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1563246_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1563246_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1563246_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1563246_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1563246_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1563246_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1563246_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1563246_at9 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A018125 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A018125 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 229254 | - | - |
