Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002903.hg.1 |
ENST00000414504.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002903.hg.1 |
ENST00000424524.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002903.hg.1 |
NR_002717.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002903.hg.1 |
TC13000264.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002902.hg.1 |
ENST00000414504.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002902.hg.1 |
ENST00000424524.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002902.hg.1 |
NR_002717.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002902.hg.1 |
TC13000264.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007446.hg.1 |
ENST00000414504.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007446.hg.1 |
ENST00000414504.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007446.hg.1 |
ENST00000414504.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007446.hg.1 |
ENST00000414504.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007446.hg.1 |
ENST00000414504.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007446.hg.1 |
ENST00000414504.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007446.hg.1 |
ENST00000414504.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007446.hg.1 |
ENST00000414504.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007446.hg.1 |
ENST00000414504.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007446.hg.1 |
ENST00000414504.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007446.hg.1 |
ENST00000414504.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007446.hg.1 |
ENST00000414504.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007446.hg.1 |
ENST00000424524.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007446.hg.1 |
ENST00000424524.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007446.hg.1 |
ENST00000424524.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007446.hg.1 |
ENST00000424524.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007446.hg.1 |
ENST00000424524.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007446.hg.1 |
ENST00000424524.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007446.hg.1 |
NR_002717.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007446.hg.1 |
NR_002717.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007446.hg.1 |
NR_002717.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007446.hg.1 |
NR_002717.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007446.hg.1 |
NR_002717.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007446.hg.1 |
NR_002717.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007446.hg.1 |
NR_002717.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007446.hg.1 |
NR_002717.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002900.hg.1 |
ENST00000414504.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002900.hg.1 |
NR_002717.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002900.hg.1 |
TC13000264.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1300150251.hg.1 |
ENST00000414504.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300150251.hg.1 |
ENST00000414504.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300150251.hg.1 |
ENST00000424524.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300150251.hg.1 |
NR_002717.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300150251.hg.1 |
ENST00000414504.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300150251.hg.1 |
ENST00000414504.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300150251.hg.1 |
ENST00000424524.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300150251.hg.1 |
NR_002717.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300150251.hg.1 |
TC1300007446.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300150250.hg.1 |
ENST00000414504.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300150250.hg.1 |
ENST00000414504.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300150250.hg.1 |
ENST00000424524.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300150250.hg.1 |
NR_002717.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300150250.hg.1 |
ENST00000414504.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300150250.hg.1 |
ENST00000414504.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300150250.hg.1 |
ENST00000424524.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300150250.hg.1 |
NR_002717.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300150250.hg.1 |
TC1300007446.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300150245.hg.1 |
ENST00000414504.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300150245.hg.1 |
ENST00000414504.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300150245.hg.1 |
NR_002717.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300150245.hg.1 |
ENST00000414504.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300150245.hg.1 |
ENST00000414504.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300150245.hg.1 |
NR_002717.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300150245.hg.1 |
TC1300007446.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300007446.hg.1 |
ENST00000414504.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007446.hg.1 |
ENST00000414504.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007446.hg.1 |
ENST00000414504.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007446.hg.1 |
ENST00000414504.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007446.hg.1 |
ENST00000414504.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007446.hg.1 |
ENST00000414504.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007446.hg.1 |
ENST00000414504.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007446.hg.1 |
ENST00000414504.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007446.hg.1 |
ENST00000414504.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007446.hg.1 |
ENST00000414504.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007446.hg.1 |
ENST00000414504.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007446.hg.1 |
ENST00000414504.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007446.hg.1 |
ENST00000424524.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007446.hg.1 |
ENST00000424524.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007446.hg.1 |
ENST00000424524.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007446.hg.1 |
ENST00000424524.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007446.hg.1 |
ENST00000424524.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007446.hg.1 |
ENST00000424524.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007446.hg.1 |
NR_002717.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007446.hg.1 |
NR_002717.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007446.hg.1 |
NR_002717.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007446.hg.1 |
NR_002717.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007446.hg.1 |
NR_002717.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007446.hg.1 |
NR_002717.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007446.hg.1 |
NR_002717.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007446.hg.1 |
NR_002717.2 |
Refseq |