Assay Details
| Gene Symbol: LINC01220 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038421.1 | 2 - 3 | 197 | 66 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000472.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09395246 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29855497 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09395219 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09395225 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09395224 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09395233 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09395220 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29855508 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09395245 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09395231 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29855496 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09395221 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29855500 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09395236 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29855509 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29855479 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29855493 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09395241 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09395242 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29855506 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09395247 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29855483 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09395214 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29855510 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09395235 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09395217 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09395212 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16717421 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09395213 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29855472 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09395229 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09395210 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29855517 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09395223 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 827310 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P20110801 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_18_P20110801 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 1563373_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005885.hg.1 | ENST00000558575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005885.hg.1 | NR_038421.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005885.hg.1 | TC14000472.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005884.hg.1 | ENST00000558575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005884.hg.1 | ENST00000561000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005884.hg.1 | NR_038421.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005884.hg.1 | TC14000472.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007708.hg.1 | ENST00000558575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007708.hg.1 | ENST00000558575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007708.hg.1 | ENST00000558575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007708.hg.1 | ENST00000558575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007708.hg.1 | ENST00000558575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007708.hg.1 | ENST00000561000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007708.hg.1 | ENST00000561000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007708.hg.1 | NR_038421.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007708.hg.1 | NR_038421.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007708.hg.1 | NR_038421.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007708.hg.1 | NR_038421.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005887.hg.1 | ENST00000558575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005887.hg.1 | NR_038421.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005887.hg.1 | TC14000472.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1400154483.hg.1 | ENST00000558575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154483.hg.1 | ENST00000561000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154483.hg.1 | NR_038421.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400154483.hg.1 | ENST00000558575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154483.hg.1 | ENST00000561000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154483.hg.1 | NR_038421.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400154483.hg.1 | TC1400007708.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400154482.hg.1 | ENST00000558575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154482.hg.1 | ENST00000561000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154482.hg.1 | NR_038421.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400154482.hg.1 | ENST00000558575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154482.hg.1 | ENST00000561000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154482.hg.1 | NR_038421.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400154482.hg.1 | TC1400007708.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400154487.hg.1 | ENST00000558575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154487.hg.1 | NR_038421.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400154487.hg.1 | ENST00000558575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154487.hg.1 | NR_038421.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400154487.hg.1 | TC1400007708.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400007708.hg.1 | ENST00000558575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007708.hg.1 | ENST00000558575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007708.hg.1 | ENST00000558575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007708.hg.1 | ENST00000558575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007708.hg.1 | ENST00000558575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007708.hg.1 | ENST00000561000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007708.hg.1 | ENST00000561000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007708.hg.1 | NR_038421.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007708.hg.1 | NR_038421.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007708.hg.1 | NR_038421.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007708.hg.1 | NR_038421.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400154485.hg.1 | ENST00000558575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154485.hg.1 | NR_038421.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400154485.hg.1 | ENST00000558575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154485.hg.1 | NR_038421.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400154485.hg.1 | TC1400007708.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 1563373_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1563373_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563373_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563373_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563373_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563373_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563373_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563373_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563373_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563373_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563373_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563373_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563373_at5 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A051623 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A051623 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 228184 | - | - |

LINC01220
Genomic Map