Assay Details
| Gene Symbol: NWD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144990.1 | NP_001138462.1 | 5 - 6 | 704 | 76 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000211.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04003116.hg.1 | TC04000211.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003116.hg.1 | ENST00000309447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003116.hg.1 | NM_001144990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003115.hg.1 | TC04000211.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003115.hg.1 | ENST00000309447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003115.hg.1 | NM_001144990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003117.hg.1 | TC04000211.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003117.hg.1 | ENST00000309447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003117.hg.1 | NM_001144990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003118.hg.1 | TC04000211.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003118.hg.1 | ENST00000309447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003118.hg.1 | NM_001144990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003120.hg.1 | TC04000211.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003120.hg.1 | ENST00000309447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003120.hg.1 | NM_001144990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003114.hg.1 | TC04000211.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003114.hg.1 | ENST00000309447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003114.hg.1 | NM_001144990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003119.hg.1 | TC04000211.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003119.hg.1 | ENST00000309447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003119.hg.1 | NM_001144990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007231.hg.1 | ENST00000309447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007231.hg.1 | ENST00000309447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007231.hg.1 | ENST00000309447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007231.hg.1 | ENST00000309447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007231.hg.1 | ENST00000309447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007231.hg.1 | ENST00000309447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007231.hg.1 | ENST00000309447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007231.hg.1 | ENST00000309447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007231.hg.1 | ENST00000309447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007231.hg.1 | NM_001144990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007231.hg.1 | NM_001144990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007231.hg.1 | NM_001144990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007231.hg.1 | NM_001144990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007231.hg.1 | NM_001144990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007231.hg.1 | NM_001144990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007231.hg.1 | NM_001144990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007231.hg.1 | NM_001144990.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400149428.hg.1 | ENST00000309447.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400149428.hg.1 | NM_001144990.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400149428.hg.1 | ENST00000309447.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400149428.hg.1 | NM_001144990.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400149428.hg.1 | TC0400007231.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400149423.hg.1 | ENST00000309447.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400149423.hg.1 | NM_001144990.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400149423.hg.1 | ENST00000309447.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400149423.hg.1 | NM_001144990.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400149423.hg.1 | TC0400007231.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400149425.hg.1 | ENST00000309447.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400149425.hg.1 | NM_001144990.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400149425.hg.1 | ENST00000309447.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400149425.hg.1 | NM_001144990.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400149425.hg.1 | TC0400007231.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400149429.hg.1 | ENST00000309447.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400149429.hg.1 | NM_001144990.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400149429.hg.1 | ENST00000309447.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400149429.hg.1 | NM_001144990.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400149429.hg.1 | TC0400007231.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400149424.hg.1 | ENST00000309447.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400149424.hg.1 | NM_001144990.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400149424.hg.1 | ENST00000309447.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400149424.hg.1 | NM_001144990.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400149424.hg.1 | TC0400007231.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400149427.hg.1 | ENST00000309447.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400149427.hg.1 | NM_001144990.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400149427.hg.1 | ENST00000309447.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400149427.hg.1 | NM_001144990.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400149427.hg.1 | TC0400007231.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400149426.hg.1 | ENST00000309447.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400149426.hg.1 | NM_001144990.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400149426.hg.1 | ENST00000309447.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400149426.hg.1 | NM_001144990.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400149426.hg.1 | TC0400007231.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400007231.hg.1 | ENST00000309447.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007231.hg.1 | ENST00000309447.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007231.hg.1 | ENST00000309447.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007231.hg.1 | ENST00000309447.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007231.hg.1 | ENST00000309447.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007231.hg.1 | ENST00000309447.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007231.hg.1 | ENST00000309447.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007231.hg.1 | ENST00000309447.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007231.hg.1 | ENST00000309447.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007231.hg.1 | NM_001144990.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007231.hg.1 | NM_001144990.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007231.hg.1 | NM_001144990.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007231.hg.1 | NM_001144990.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007231.hg.1 | NM_001144990.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007231.hg.1 | NM_001144990.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007231.hg.1 | NM_001144990.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007231.hg.1 | NM_001144990.1 | Refseq |

NWD2
Genomic Map