Assay Details
| Gene Symbol: LOC107984678 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC035345.1 | - | 1 - 2 | 437 | 76 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 4710 | MIR4710 | mir-4710 | -3.4kb | Anti-sense | Chr. 14 - 104677694 - 104677749 on NCBI Build GRCh38 |
| Downstream |
inverted formin, FH2 and WH2 domain containing |
INF2 |
C14orf151, C14orf173, CMTDIE, FSGS5, pp9484 |
5.3kb | Sense | Chr. 14 - 104689606 - 104719610 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC107984678
Genomic Map