Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008555.hg.1 |
ENST00000319682.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008555.hg.1 |
ENST00000319682.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008555.hg.1 |
ENST00000319682.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008555.hg.1 |
ENST00000319682.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008555.hg.1 |
ENST00000319682.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008555.hg.1 |
ENST00000319682.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008555.hg.1 |
ENST00000319682.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008555.hg.1 |
ENST00000319682.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008555.hg.1 |
NR_024564.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008555.hg.1 |
NR_024564.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008555.hg.1 |
NR_024564.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008555.hg.1 |
NR_024564.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008555.hg.1 |
NR_024564.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008555.hg.1 |
NR_024564.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008555.hg.1 |
NR_024564.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008555.hg.1 |
NR_024564.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009430.hg.1 |
ENST00000319682.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009430.hg.1 |
NR_024564.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009430.hg.1 |
TC20000675.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009431.hg.1 |
ENST00000319682.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009431.hg.1 |
NR_024564.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009431.hg.1 |
TC20000675.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009432.hg.1 |
ENST00000319682.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009432.hg.1 |
NR_024564.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009432.hg.1 |
TC20000675.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000160045.hg.1 |
ENST00000319682.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160045.hg.1 |
NR_024564.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160045.hg.1 |
TC2000008555.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000160045.hg.1 |
ENST00000319682.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160045.hg.1 |
NR_024564.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008555.hg.1 |
ENST00000319682.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008555.hg.1 |
ENST00000319682.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008555.hg.1 |
ENST00000319682.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008555.hg.1 |
ENST00000319682.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008555.hg.1 |
ENST00000319682.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008555.hg.1 |
ENST00000319682.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008555.hg.1 |
ENST00000319682.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008555.hg.1 |
ENST00000319682.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008555.hg.1 |
NR_024564.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008555.hg.1 |
NR_024564.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008555.hg.1 |
NR_024564.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008555.hg.1 |
NR_024564.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008555.hg.1 |
NR_024564.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008555.hg.1 |
NR_024564.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008555.hg.1 |
NR_024564.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008555.hg.1 |
NR_024564.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160047.hg.1 |
ENST00000319682.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160047.hg.1 |
NR_024564.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160047.hg.1 |
TC2000008555.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000160047.hg.1 |
ENST00000319682.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160047.hg.1 |
NR_024564.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160046.hg.1 |
ENST00000319682.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160046.hg.1 |
NR_024564.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160046.hg.1 |
TC2000008555.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000160046.hg.1 |
ENST00000319682.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160046.hg.1 |
NR_024564.1 |
Refseq |