Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008387.hg.1 |
ENST00000448859.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008387.hg.1 |
ENST00000448859.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008387.hg.1 |
ENST00000448859.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008387.hg.1 |
ENST00000448859.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008387.hg.1 |
ENST00000448859.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008387.hg.1 |
ENST00000448859.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008387.hg.1 |
ENST00000448859.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008387.hg.1 |
ENST00000448859.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008387.hg.1 |
NR_109866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008387.hg.1 |
NR_109866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008387.hg.1 |
NR_109866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008387.hg.1 |
NR_109866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008387.hg.1 |
NR_109866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008387.hg.1 |
NR_109866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008387.hg.1 |
NR_109866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008387.hg.1 |
NR_109866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000159065.hg.1 |
ENST00000448859.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000159065.hg.1 |
NR_109866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000159065.hg.1 |
TC2000008387.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000159065.hg.1 |
ENST00000448859.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000159065.hg.1 |
NR_109866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008387.hg.1 |
ENST00000448859.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008387.hg.1 |
ENST00000448859.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008387.hg.1 |
ENST00000448859.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008387.hg.1 |
ENST00000448859.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008387.hg.1 |
ENST00000448859.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008387.hg.1 |
ENST00000448859.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008387.hg.1 |
ENST00000448859.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008387.hg.1 |
ENST00000448859.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008387.hg.1 |
NR_109866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008387.hg.1 |
NR_109866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008387.hg.1 |
NR_109866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008387.hg.1 |
NR_109866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008387.hg.1 |
NR_109866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008387.hg.1 |
NR_109866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008387.hg.1 |
NR_109866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008387.hg.1 |
NR_109866.1 |
Refseq |