Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007700.hg.1 |
ENST00000426840.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007700.hg.1 |
ENST00000426840.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007700.hg.1 |
ENST00000426840.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007700.hg.1 |
ENST00000426840.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007700.hg.1 |
ENST00000426840.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007700.hg.1 |
ENST00000426840.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007700.hg.1 |
NR_120414.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007700.hg.1 |
NR_120414.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007700.hg.1 |
NR_120414.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007700.hg.1 |
NR_120414.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007700.hg.1 |
NR_120414.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007700.hg.1 |
NR_120414.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007700.hg.1 |
NR_120414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300151945.hg.1 |
ENST00000426840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300151945.hg.1 |
NR_120414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300151945.hg.1 |
ENST00000426840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300151945.hg.1 |
NR_120414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300151945.hg.1 |
TC1300007700.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300007700.hg.1 |
ENST00000426840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007700.hg.1 |
ENST00000426840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007700.hg.1 |
ENST00000426840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007700.hg.1 |
ENST00000426840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007700.hg.1 |
ENST00000426840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007700.hg.1 |
ENST00000426840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007700.hg.1 |
NR_120414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007700.hg.1 |
NR_120414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007700.hg.1 |
NR_120414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007700.hg.1 |
NR_120414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007700.hg.1 |
NR_120414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007700.hg.1 |
NR_120414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007700.hg.1 |
NR_120414.1 |
Refseq |