| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007700.hg.1 |
ENST00000426840.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007700.hg.1 |
ENST00000426840.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007700.hg.1 |
ENST00000426840.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007700.hg.1 |
ENST00000426840.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007700.hg.1 |
ENST00000426840.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007700.hg.1 |
ENST00000426840.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007700.hg.1 |
NR_120414.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007700.hg.1 |
NR_120414.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007700.hg.1 |
NR_120414.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007700.hg.1 |
NR_120414.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007700.hg.1 |
NR_120414.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007700.hg.1 |
NR_120414.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007700.hg.1 |
NR_120414.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007700.hg.1 |
ENST00000426840.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007700.hg.1 |
ENST00000426840.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007700.hg.1 |
ENST00000426840.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007700.hg.1 |
ENST00000426840.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007700.hg.1 |
ENST00000426840.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007700.hg.1 |
ENST00000426840.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007700.hg.1 |
NR_120414.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007700.hg.1 |
NR_120414.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007700.hg.1 |
NR_120414.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007700.hg.1 |
NR_120414.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007700.hg.1 |
NR_120414.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007700.hg.1 |
NR_120414.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007700.hg.1 |
NR_120414.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300151947.hg.1 |
ENST00000426840.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300151947.hg.1 |
NR_120414.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300151947.hg.1 |
ENST00000426840.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300151947.hg.1 |
NR_120414.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300151947.hg.1 |
TC1300007700.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300151948.hg.1 |
NR_120414.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300151948.hg.1 |
NR_120414.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300151948.hg.1 |
TC1300007700.hg.1 |
Clariom D TC |