Assay Details
Gene Symbol: MIR17HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_027349.1 | 2 - 3 | 210 | 83 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 379 | LINC00379 | -138.0kb | Anti-sense | Chr. 13 - 91127613 - 91211698 on NCBI Build GRCh38 | |
Downstream | microRNA 17 | MIR17 |
MIR17-5p, MIR91, MIRN17, MIRN91, hsa-mir-17, miR-17, miR17-3p, miRNA17, miRNA91 |
379.0 | Sense | Chr. 13 - 91350605 - 91350688 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC13000322.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR13003381.hg.1 | ENST00000400282.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13003381.hg.1 | NR_027349.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13003381.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13003381.hg.1 | TC13000322.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000362279.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000362310.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000384829.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000384878.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000385012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000385233.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000400282.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000400282.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000400282.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000400282.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000400282.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000400282.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000400282.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000400282.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000581816.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000581816.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000581816.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000581816.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000581816.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000581816.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000581816.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000581816.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000581816.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000581816.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000581816.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000581816.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000581816.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000581816.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000581816.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | NR_027349.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | NR_027349.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | NR_027349.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | NR_027349.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | NR_029487.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | NR_029488.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | NR_029489.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | NR_029490.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | NR_029492.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007706.hg.1 | NR_029508.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000362279.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000362310.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000384829.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000384878.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000385012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000385233.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000400282.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000400282.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000400282.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000400282.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000400282.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000400282.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000400282.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000400282.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000581816.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000581816.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000581816.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000581816.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000581816.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000581816.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000581816.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000581816.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000581816.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000581816.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000581816.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000581816.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000581816.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000581816.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000581816.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | NR_027349.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | NR_027349.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | NR_027349.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | NR_027349.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | NR_029487.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | NR_029488.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | NR_029489.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | NR_029490.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | NR_029492.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007706.hg.1 | NR_029508.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300152009.hg.1 | ENST00000400282.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300152009.hg.1 | ENST00000400282.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300152009.hg.1 | ENST00000581816.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300152009.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300152009.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300152009.hg.1 | NR_027349.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300152009.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300152009.hg.1 | ENST00000400282.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300152009.hg.1 | ENST00000400282.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300152009.hg.1 | ENST00000581816.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300152009.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300152009.hg.1 | ENST00000582141.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300152009.hg.1 | NR_027349.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300152009.hg.1 | NR_027350.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300152009.hg.1 | TC1300007706.hg.1 | Clariom D TC |
