Assay Details
| Gene Symbol: TEPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199046.2 | NP_950247.2 | 6 - 7 | 872 | 73 | |
| NM_199456.2 | NP_955535.2 | 6 - 7 | 791 | 73 | ||
| GenBank mRNA | AY363222.1 | - | 6 - 7 | 791 | 73 | |
| AY363223.1 | - | 6 - 7 | 872 | 73 | ||
| BC104458.1 | - | 6 - 7 | 782 | 40030655 | 73 | |
| BC104459.1 | - | 6 - 7 | 782 | 40030657 | 73 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000498.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000290871.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000290871.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000290871.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000290871.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000290871.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000290871.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000290871.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000290871.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000290871.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000290871.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000290871.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000290871.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000290871.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000441824.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000441824.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000441824.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000441824.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000441824.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000441824.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000441824.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000441824.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000441824.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000441824.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000441824.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000441824.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000562915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000562915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000562915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000562915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000562915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000562915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000562915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000562915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000562915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000562915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000562915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000569996.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000569996.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000569996.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000569996.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000569996.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000569996.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000569996.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000569996.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | ENST00000569996.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | NM_199046.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | NM_199046.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | NM_199046.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | NM_199046.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | NM_199046.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | NM_199046.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | NM_199046.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | NM_199046.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | NM_199046.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | NM_199046.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | NM_199046.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | NM_199046.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | NM_199046.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | NM_199046.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | NM_199456.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | NM_199456.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | NM_199456.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | NM_199456.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | NM_199456.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | NM_199456.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | NM_199456.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | NM_199456.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | NM_199456.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | NM_199456.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | NM_199456.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | NM_199456.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008019.hg.1 | NM_199456.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16006969.hg.1 | ENST00000290871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16006969.hg.1 | ENST00000441824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16006969.hg.1 | NM_199046.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16006969.hg.1 | NM_199456.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16006969.hg.1 | TC16000498.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000290871.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000290871.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000290871.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000290871.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000290871.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000290871.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000290871.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000290871.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000290871.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000290871.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000290871.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000290871.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000290871.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000441824.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000441824.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000441824.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000441824.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000441824.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000441824.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000441824.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000441824.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000441824.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000441824.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000441824.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000441824.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000562915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000562915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000562915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000562915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000562915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000562915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000562915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000562915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000562915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000562915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000562915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000569996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000569996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000569996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000569996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000569996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000569996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000569996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000569996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | ENST00000569996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | NM_199046.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | NM_199046.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | NM_199046.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | NM_199046.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | NM_199046.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | NM_199046.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | NM_199046.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | NM_199046.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | NM_199046.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | NM_199046.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | NM_199046.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | NM_199046.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | NM_199046.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | NM_199046.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | NM_199456.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | NM_199456.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | NM_199456.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | NM_199456.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | NM_199456.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | NM_199456.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | NM_199456.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | NM_199456.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | NM_199456.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | NM_199456.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | NM_199456.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | NM_199456.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008019.hg.1 | NM_199456.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600159136.hg.1 | ENST00000290871.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600159136.hg.1 | ENST00000441824.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600159136.hg.1 | ENST00000562915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600159136.hg.1 | ENST00000569996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600159136.hg.1 | NM_199046.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600159136.hg.1 | NM_199456.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600159136.hg.1 | ENST00000290871.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600159136.hg.1 | ENST00000441824.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600159136.hg.1 | ENST00000562915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600159136.hg.1 | ENST00000569996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600159136.hg.1 | NM_199046.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600159136.hg.1 | NM_199456.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600159136.hg.1 | TC1600008019.hg.1 | Clariom D TC |

TEPP
Genomic Map