Assay Details
Gene Symbol: LOC100133669 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_026913.1 | 1 - 2 | 231 | 67 | ||
GenBank mRNA | BM557790.1 | - | 1 - 2 | 231 | 67 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | cell division cycle 42 pseudogene 3 | CDC42P3 | Anti-sense | Chr. 8 - 142995828 - 142997663 on NCBI Build GRCh38 | ||
Downstream |
lymphocyte antigen 6 complex, locus E |
LY6E |
RIG-E, RIGE, SCA-2, SCA2, TSA-1 |
95.0 | Anti-sense | Chr. 8 - 143018485 - 143022410 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human CGH Microarray 4x180K | 13403 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 140986 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 275760 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 461819 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 552973 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 668207 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 860545 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 777634 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 641062 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 133078 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 9661 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 5630 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 66189 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 822977 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 5339 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 349916 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3251492 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4847 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4846 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 14482 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P02042304 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P59966906 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P38615611 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P38615611 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P59966906 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3251492 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3251492 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06531101 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06531099 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26815394 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26815392 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06531100 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06531108 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06531089 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16145553 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06531104 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06531095 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06531102 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06531103 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26815426 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06531107 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26815397 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16145554 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26815417 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06531106 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06531091 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16145554 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26815392 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26815397 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06531099 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06531108 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06531104 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06531100 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26815426 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26815417 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06531107 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06531101 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26815394 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06531095 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06531106 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06531091 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06531102 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06531103 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06531089 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16145553 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P18519994 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02042304 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02042307 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06531084 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06531028 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06531031 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16145494 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06531093 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06531088 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06531103 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06531108 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06531082 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06531056 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06531058 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06531032 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06531074 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06531089 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06531080 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06531070 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06531095 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06531104 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06531038 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06531106 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06531067 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06531033 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06531085 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06531052 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06531078 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16145507 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06531054 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16145509 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16145491 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06531072 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06531086 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06531043 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06531026 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06531048 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06531036 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06531064 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06531040 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06531060 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06531041 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06531099 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06531092 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06531063 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06531101 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06531050 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06531076 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P02042298 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P02042304 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P38615611 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P18519957 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 1561850_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC0800012097.hg.1 | ENST00000502167.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012097.hg.1 | ENST00000502167.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012097.hg.1 | ENST00000502167.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012097.hg.1 | ENST00000502167.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012097.hg.1 | ENST00000517833.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012097.hg.1 | ENST00000517833.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012097.hg.1 | ENST00000517833.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012097.hg.1 | ENST00000517833.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012097.hg.1 | ENST00000518024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012097.hg.1 | ENST00000518024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012097.hg.1 | ENST00000518831.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012097.hg.1 | ENST00000518831.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012097.hg.1 | ENST00000518831.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012097.hg.1 | ENST00000522060.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012097.hg.1 | ENST00000522060.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012097.hg.1 | ENST00000522060.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08022493.hg.1 | TC08001710.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08022493.hg.1 | NR_026913.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08022501.hg.1 | TC08001710.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08022501.hg.1 | ENST00000502167.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08022501.hg.1 | NR_026913.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012097.hg.1 | ENST00000502167.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012097.hg.1 | ENST00000502167.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012097.hg.1 | ENST00000502167.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012097.hg.1 | ENST00000502167.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012097.hg.1 | ENST00000517833.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012097.hg.1 | ENST00000517833.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012097.hg.1 | ENST00000517833.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012097.hg.1 | ENST00000517833.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012097.hg.1 | ENST00000518024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012097.hg.1 | ENST00000518024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012097.hg.1 | ENST00000518831.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012097.hg.1 | ENST00000518831.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012097.hg.1 | ENST00000518831.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012097.hg.1 | ENST00000522060.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012097.hg.1 | ENST00000522060.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012097.hg.1 | ENST00000522060.1 | Ensembl |
Human Genome U133 Plus 2.0 Array | 1561850_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 1561850_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1561850_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1561850_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1561850_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1561850_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1561850_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1561850_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1561850_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1561850_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1561850_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1561850_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1561850_at10 | - | - |
Human Genome U95 Set | 55115_at | - | - |
Human Genome U95 Set | 55115_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A036203 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A036203 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 232711 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3203238 | BM557790.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3203238 | NR_026913.1 | human_transcriptome |
