| Array Name | 
									Probe IDs | 
									Targets | 
									Target Type | 
								
	
								
									
										
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC0700009790.hg.1 | 
												ENST00000397551.6 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC0700009790.hg.1 | 
												ENST00000397551.6 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC0700009790.hg.1 | 
												ENST00000397551.6 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC0700009790.hg.1 | 
												ENST00000397551.6 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC0700009790.hg.1 | 
												ENST00000411526.5 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC0700009790.hg.1 | 
												ENST00000411526.5 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC0700009790.hg.1 | 
												ENST00000411526.5 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC0700009790.hg.1 | 
												ENST00000411526.5 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC0700009790.hg.1 | 
												ENST00000411526.5 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC0700009790.hg.1 | 
												ENST00000411526.5 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC0700009790.hg.1 | 
												ENST00000449486.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC0700009790.hg.1 | 
												ENST00000449486.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC0700009790.hg.1 | 
												ENST00000449486.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC0700009790.hg.1 | 
												ENST00000449486.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC0700009790.hg.1 | 
												ENST00000449486.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC0700009790.hg.1 | 
												ENST00000449486.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC0700009790.hg.1 | 
												ENST00000449486.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC0700009790.hg.1 | 
												ENST00000449486.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC0700009790.hg.1 | 
												ENST00000449486.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC0700009790.hg.1 | 
												ENST00000449486.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC0700009790.hg.1 | 
												ENST00000449486.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC0700009790.hg.1 | 
												NR_024476.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC0700009790.hg.1 | 
												NR_024476.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC0700009790.hg.1 | 
												NR_024476.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC0700009790.hg.1 | 
												NR_024476.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC0700009790.hg.1 | 
												NR_024476.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC0700009790.hg.1 | 
												NR_024476.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC0700009790.hg.1 | 
												NR_024476.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC0700009790.hg.1 | 
												NR_024476.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC0700009790.hg.1 | 
												NR_024476.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC0700009790.hg.1 | 
												NR_024476.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC0700009790.hg.1 | 
												NR_024477.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC0700009790.hg.1 | 
												NR_024477.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC0700009790.hg.1 | 
												NR_024477.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC0700009790.hg.1 | 
												NR_024477.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC0700009790.hg.1 | 
												NR_024477.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC0700009790.hg.1 | 
												NR_024477.1 | 
												Refseq | 
												
											
										
										
									
								
									
										
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												PSR07016018.hg.1 | 
												TC07001033.hg.1 | 
												HTA TC | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												PSR07016018.hg.1 | 
												ENST00000397551.2 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												PSR07016018.hg.1 | 
												NR_024476.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												PSR07016018.hg.1 | 
												NR_024477.1 | 
												Refseq | 
												
											
										
										
									
								
									
										
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												PSR07016019.hg.1 | 
												TC07001033.hg.1 | 
												HTA TC | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												PSR07016019.hg.1 | 
												ENST00000397551.2 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												PSR07016019.hg.1 | 
												NR_024476.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												PSR07016019.hg.1 | 
												NR_024477.1 | 
												Refseq | 
												
											
										
										
									
								
									
										
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR0700169727.hg.1 | 
												ENST00000397551.6 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR0700169727.hg.1 | 
												ENST00000411526.5 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR0700169727.hg.1 | 
												ENST00000449486.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR0700169727.hg.1 | 
												NR_024476.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR0700169727.hg.1 | 
												NR_024477.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR0700169727.hg.1 | 
												ENST00000397551.6 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR0700169727.hg.1 | 
												ENST00000411526.5 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR0700169727.hg.1 | 
												ENST00000449486.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR0700169727.hg.1 | 
												NR_024476.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR0700169727.hg.1 | 
												NR_024477.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR0700169727.hg.1 | 
												TC0700009790.hg.1 | 
												Clariom D TC | 
												
											
										
										
									
								
									
										
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR0700169726.hg.1 | 
												ENST00000397551.6 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR0700169726.hg.1 | 
												ENST00000411526.5 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR0700169726.hg.1 | 
												ENST00000449486.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR0700169726.hg.1 | 
												NR_024476.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR0700169726.hg.1 | 
												NR_024477.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR0700169726.hg.1 | 
												ENST00000397551.6 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR0700169726.hg.1 | 
												ENST00000411526.5 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR0700169726.hg.1 | 
												ENST00000449486.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR0700169726.hg.1 | 
												NR_024476.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR0700169726.hg.1 | 
												NR_024477.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR0700169726.hg.1 | 
												TC0700009790.hg.1 | 
												Clariom D TC | 
												
											
										
										
									
								
									
										
											
												
												| Affymetrix Human Clariom D Assay | 
												TC0700009790.hg.1 | 
												ENST00000397551.6 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC0700009790.hg.1 | 
												ENST00000397551.6 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC0700009790.hg.1 | 
												ENST00000397551.6 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC0700009790.hg.1 | 
												ENST00000397551.6 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC0700009790.hg.1 | 
												ENST00000411526.5 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC0700009790.hg.1 | 
												ENST00000411526.5 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC0700009790.hg.1 | 
												ENST00000411526.5 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC0700009790.hg.1 | 
												ENST00000411526.5 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC0700009790.hg.1 | 
												ENST00000411526.5 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC0700009790.hg.1 | 
												ENST00000411526.5 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC0700009790.hg.1 | 
												ENST00000449486.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC0700009790.hg.1 | 
												ENST00000449486.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC0700009790.hg.1 | 
												ENST00000449486.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC0700009790.hg.1 | 
												ENST00000449486.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC0700009790.hg.1 | 
												ENST00000449486.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC0700009790.hg.1 | 
												ENST00000449486.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC0700009790.hg.1 | 
												ENST00000449486.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC0700009790.hg.1 | 
												ENST00000449486.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC0700009790.hg.1 | 
												ENST00000449486.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC0700009790.hg.1 | 
												ENST00000449486.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC0700009790.hg.1 | 
												ENST00000449486.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC0700009790.hg.1 | 
												NR_024476.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC0700009790.hg.1 | 
												NR_024476.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC0700009790.hg.1 | 
												NR_024476.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC0700009790.hg.1 | 
												NR_024476.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC0700009790.hg.1 | 
												NR_024476.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC0700009790.hg.1 | 
												NR_024476.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC0700009790.hg.1 | 
												NR_024476.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC0700009790.hg.1 | 
												NR_024476.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC0700009790.hg.1 | 
												NR_024476.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC0700009790.hg.1 | 
												NR_024476.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC0700009790.hg.1 | 
												NR_024477.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC0700009790.hg.1 | 
												NR_024477.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC0700009790.hg.1 | 
												NR_024477.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC0700009790.hg.1 | 
												NR_024477.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC0700009790.hg.1 | 
												NR_024477.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC0700009790.hg.1 | 
												NR_024477.1 | 
												Refseq |