Assay Details
| Gene Symbol: MGC72080 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002822.3 | 3 - 4 | 309 | 79 | ||
| GenBank mRNA | BC029615.1 | - | 3 - 4 | 319 | 79 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001631.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25890081 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 22567 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 8x60K | 19673 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 85537 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 63583 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 383500 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 262008 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 209359 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 380368 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 360637 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 249071 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3280400 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3280418 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3900 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 8680 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 15887 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P01762679 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P01762679 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3213557 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3280400 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3280418 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3280400 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3280418 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25890081 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05638693 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05638695 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25890116 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05638691 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15992443 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15992473 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25890084 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25890101 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05638690 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05638690 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25890084 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25890101 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05638691 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25890116 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05638693 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05638695 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25890081 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15992443 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15992473 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05638688 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05638691 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05638692 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05638687 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05638696 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P01762679 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P01762680 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NR_002822_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7968788 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8045888 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8141170 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8141171 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8042575 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8082249 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7971242 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8138152 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7945874 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8156320 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8082245 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7938684 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8138146 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8141169 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8042574 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7968787 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8156319 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8045887 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7945873 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8082248 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8138151 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7971241 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8138145 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8082244 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR07024827.hg.1 | TC07001631.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07024827.hg.1 | NR_002822.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07024825.hg.1 | TC07001631.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07024825.hg.1 | NR_002822.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07024823.hg.1 | TC07001631.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07024823.hg.1 | NR_002822.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07024824.hg.1 | TC07001631.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07024824.hg.1 | NR_002822.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011877.hg.1 | ENST00000442908.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011877.hg.1 | ENST00000442908.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011877.hg.1 | ENST00000442908.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011877.hg.1 | ENST00000442908.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011877.hg.1 | ENST00000442908.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011877.hg.1 | ENST00000442908.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011877.hg.1 | ENST00000442908.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011877.hg.1 | ENST00000442908.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011877.hg.1 | ENST00000442908.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011877.hg.1 | ENST00000442908.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011877.hg.1 | ENST00000442908.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011877.hg.1 | ENST00000453589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011877.hg.1 | ENST00000453589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011877.hg.1 | ENST00000453589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011877.hg.1 | ENST00000453589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011877.hg.1 | ENST00000453589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011877.hg.1 | ENST00000453589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011877.hg.1 | ENST00000453589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011877.hg.1 | NR_002822.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011877.hg.1 | NR_002822.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011877.hg.1 | NR_002822.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011877.hg.1 | NR_002822.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011877.hg.1 | NR_002822.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011877.hg.1 | NR_002822.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011877.hg.1 | NR_002822.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700185009.hg.1 | ENST00000453589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700185009.hg.1 | NR_002822.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700185009.hg.1 | ENST00000453589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700185009.hg.1 | NR_002822.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700185009.hg.1 | TC0700011877.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700185013.hg.1 | ENST00000453589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700185013.hg.1 | NR_002822.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700185013.hg.1 | ENST00000453589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700185013.hg.1 | NR_002822.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700185013.hg.1 | TC0700011877.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700185039.hg.1 | NR_002822.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700185039.hg.1 | NR_002822.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700185039.hg.1 | TC0700011877.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700185037.hg.1 | ENST00000453589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700185037.hg.1 | NR_002822.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700185037.hg.1 | ENST00000453589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700185037.hg.1 | NR_002822.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700185037.hg.1 | TC0700011877.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700011877.hg.1 | ENST00000442908.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011877.hg.1 | ENST00000442908.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011877.hg.1 | ENST00000442908.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011877.hg.1 | ENST00000442908.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011877.hg.1 | ENST00000442908.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011877.hg.1 | ENST00000442908.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011877.hg.1 | ENST00000442908.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011877.hg.1 | ENST00000442908.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011877.hg.1 | ENST00000442908.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011877.hg.1 | ENST00000442908.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011877.hg.1 | ENST00000442908.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011877.hg.1 | ENST00000453589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011877.hg.1 | ENST00000453589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011877.hg.1 | ENST00000453589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011877.hg.1 | ENST00000453589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011877.hg.1 | ENST00000453589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011877.hg.1 | ENST00000453589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011877.hg.1 | ENST00000453589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011877.hg.1 | NR_002822.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011877.hg.1 | NR_002822.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011877.hg.1 | NR_002822.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011877.hg.1 | NR_002822.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011877.hg.1 | NR_002822.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011877.hg.1 | NR_002822.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011877.hg.1 | NR_002822.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700185002.hg.1 | ENST00000442908.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700185002.hg.1 | NR_002822.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700185002.hg.1 | ENST00000442908.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700185002.hg.1 | NR_002822.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700185002.hg.1 | TC0700011877.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700185036.hg.1 | ENST00000453589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700185036.hg.1 | NR_002822.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700185036.hg.1 | ENST00000453589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700185036.hg.1 | NR_002822.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700185036.hg.1 | TC0700011877.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700185038.hg.1 | NR_002822.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700185038.hg.1 | NR_002822.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700185038.hg.1 | TC0700011877.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A033200 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A033200 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 143725 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_2152502 | NR_002822.3 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2152502 | BC015431.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2152502 | BC029615.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2152502 | NR_002822.3 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2152502 | BC015431.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2152502 | BC029615.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2152502 | NR_002822.3 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2152502 | BC015431.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2152502 | BC029615.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2152502 | NR_002822.3 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2152502 | BC015431.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2152502 | BC029615.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2152502 | NR_002822.3 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2152502 | BC015431.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2152502 | BC029615.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2152502 | NR_002822.3 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2152502 | BC015431.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2152502 | BC029615.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2152502 | NR_002822.3 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2152502 | BC015431.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2152502 | BC029615.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2152502 | NR_002822.3 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2152502 | BC015431.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2152502 | BC029615.1 | human_transcriptome |

MGC72080
Genomic Map