Assay Details
Gene Symbol: LOC101927550 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110102.1 | 4 - 5 | 66 | |||
GenBank mRNA | AI805189.1 | - | 1 - 2 | 62 | 66 | |
AK001533.1 | - | 4 - 5 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
