Assay Details
| Gene Symbol: RAET1E-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045126.1 | 1 - 2 | 883 | 93 | ||
| GenBank mRNA | BC016788.1 | - | 1 - 2 | 93 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001086.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | ENST00000446954.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | ENST00000446954.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | ENST00000446954.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | ENST00000446954.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | ENST00000596229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | ENST00000596229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | ENST00000596229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | ENST00000596229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | ENST00000596229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | ENST00000605899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | ENST00000605899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | ENST00000605899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | ENST00000605899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | ENST00000606915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | ENST00000606915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | ENST00000606915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | ENST00000606915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | ENST00000606915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | ENST00000606915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | ENST00000606915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | NR_045126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | NR_045126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | NR_045126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | NR_045126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | NR_045126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | NR_045126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | NR_045126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | NR_045127.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | NR_045127.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | NR_045127.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | NR_045127.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06012847.hg.1 | TC06001086.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06012847.hg.1 | NR_045126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06012851.hg.1 | TC06001086.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06012851.hg.1 | ENST00000446954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06012851.hg.1 | NR_045126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06012851.hg.1 | NR_045127.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600167197.hg.1 | ENST00000446954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600167197.hg.1 | ENST00000605899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600167197.hg.1 | ENST00000606915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600167197.hg.1 | NR_045126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600167197.hg.1 | NR_045127.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600167197.hg.1 | ENST00000446954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600167197.hg.1 | ENST00000605899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600167197.hg.1 | ENST00000606915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600167197.hg.1 | NR_045126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600167197.hg.1 | NR_045127.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600167197.hg.1 | TC0600009813.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600167184.hg.1 | ENST00000596229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600167184.hg.1 | ENST00000606915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600167184.hg.1 | NR_045126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600167184.hg.1 | ENST00000596229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600167184.hg.1 | ENST00000606915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600167184.hg.1 | NR_045126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600167184.hg.1 | TC0600009813.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600167186.hg.1 | ENST00000606915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600167186.hg.1 | NR_045126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600167186.hg.1 | ENST00000606915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600167186.hg.1 | NR_045126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600167186.hg.1 | TC0600009813.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600167182.hg.1 | ENST00000596229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600167182.hg.1 | ENST00000606915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600167182.hg.1 | NR_045126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600167182.hg.1 | ENST00000596229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600167182.hg.1 | ENST00000606915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600167182.hg.1 | NR_045126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600167182.hg.1 | TC0600009813.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600167183.hg.1 | ENST00000596229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600167183.hg.1 | ENST00000606915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600167183.hg.1 | NR_045126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600167183.hg.1 | ENST00000596229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600167183.hg.1 | ENST00000606915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600167183.hg.1 | NR_045126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600167183.hg.1 | TC0600009813.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600167181.hg.1 | ENST00000596229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600167181.hg.1 | ENST00000606915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600167181.hg.1 | NR_045126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600167181.hg.1 | ENST00000596229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600167181.hg.1 | ENST00000606915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600167181.hg.1 | NR_045126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600167181.hg.1 | TC0600009813.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600167196.hg.1 | ENST00000446954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600167196.hg.1 | ENST00000605899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600167196.hg.1 | ENST00000606915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600167196.hg.1 | NR_045126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600167196.hg.1 | NR_045127.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600167196.hg.1 | ENST00000446954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600167196.hg.1 | ENST00000605899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600167196.hg.1 | ENST00000606915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600167196.hg.1 | NR_045126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600167196.hg.1 | NR_045127.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600167196.hg.1 | TC0600009813.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | ENST00000446954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | ENST00000446954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | ENST00000446954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | ENST00000446954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | ENST00000596229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | ENST00000596229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | ENST00000596229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | ENST00000596229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | ENST00000596229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | ENST00000605899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | ENST00000605899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | ENST00000605899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | ENST00000605899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | ENST00000606915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | ENST00000606915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | ENST00000606915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | ENST00000606915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | ENST00000606915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | ENST00000606915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | ENST00000606915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | NR_045126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | NR_045126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | NR_045126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | NR_045126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | NR_045126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | NR_045126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | NR_045126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | NR_045127.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | NR_045127.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | NR_045127.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | NR_045127.1 | Refseq |

RAET1E-AS1
Genomic Map