Assay Details
| Gene Symbol: CENPW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012507.3 | NP_001012525.1 | 1 - 2 | 613 | 85 | |
| NR_104462.1 | 1 - 2 | 613 | 85 | |||
| GenBank mRNA | AL832235.1 | - | 1 - 2 | 613 | 85 | |
| AY902475.1 | - | 1 - 2 | 282 | 85 | ||
| BC017928.1 | - | 1 - 2 | 230 | 4655418 | 85 | |
| BC039556.2 | - | 1 - 2 | 295 | 4452583 | 85 | |
| BC046178.2 | - | 1 - 2 | 245 | 4448513 | 85 | |
| BC062798.1 | - | 1 - 2 | 258 | 6495746 | 85 | |
| BM721156.1 | - | 1 - 2 | 232 | 85 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000966.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06011308.hg.1 | TC06000966.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011308.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011313.hg.1 | TC06000966.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011313.hg.1 | ENST00000368325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011313.hg.1 | ENST00000368328.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011313.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | ENST00000368325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | ENST00000368325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | ENST00000368325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | ENST00000368325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | ENST00000368325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | ENST00000368325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | ENST00000368326.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | ENST00000368326.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | ENST00000368326.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | ENST00000368326.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | ENST00000368328.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | ENST00000368328.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | ENST00000368328.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | ENST00000368328.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | ENST00000368328.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | ENST00000368328.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001286525.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001286525.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001286525.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001286525.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001286525.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001286525.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001286525.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011314.hg.1 | TC06000966.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011314.hg.1 | ENST00000368325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011314.hg.1 | ENST00000368326.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011314.hg.1 | ENST00000368328.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011314.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011311.hg.1 | TC06000966.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011311.hg.1 | ENST00000368325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011311.hg.1 | ENST00000368326.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011311.hg.1 | ENST00000368328.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011311.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011315.hg.1 | TC06000966.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011315.hg.1 | ENST00000368325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011315.hg.1 | ENST00000368326.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011315.hg.1 | ENST00000368328.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011315.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007182.hg.1 | ENST00000553515.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007182.hg.1 | ENST00000553515.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600164373.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164373.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164373.hg.1 | NM_001286525.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164373.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164373.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164373.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164373.hg.1 | NM_001286525.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164373.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164373.hg.1 | TC0600009381.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600164374.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164374.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164374.hg.1 | NM_001286525.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164374.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164374.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164374.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164374.hg.1 | NM_001286525.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164374.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164374.hg.1 | TC0600009381.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600164377.hg.1 | ENST00000368325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600164377.hg.1 | ENST00000368326.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600164377.hg.1 | ENST00000368328.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600164377.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164377.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164377.hg.1 | NM_001286525.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164377.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164377.hg.1 | ENST00000368325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600164377.hg.1 | ENST00000368326.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600164377.hg.1 | ENST00000368328.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600164377.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164377.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164377.hg.1 | NM_001286525.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164377.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164377.hg.1 | TC0600009381.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600164378.hg.1 | ENST00000368325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600164378.hg.1 | ENST00000368326.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600164378.hg.1 | ENST00000368328.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600164378.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164378.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164378.hg.1 | NM_001286525.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164378.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164378.hg.1 | ENST00000368325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600164378.hg.1 | ENST00000368326.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600164378.hg.1 | ENST00000368328.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600164378.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164378.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164378.hg.1 | NM_001286525.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164378.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164378.hg.1 | TC0600009381.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | ENST00000368325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | ENST00000368325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | ENST00000368325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | ENST00000368325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | ENST00000368325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | ENST00000368325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | ENST00000368326.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | ENST00000368326.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | ENST00000368326.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | ENST00000368326.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | ENST00000368328.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | ENST00000368328.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | ENST00000368328.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | ENST00000368328.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | ENST00000368328.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | ENST00000368328.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001286525.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001286525.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001286525.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001286525.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001286525.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001286525.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001286525.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007182.hg.1 | ENST00000553515.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007182.hg.1 | ENST00000553515.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600164380.hg.1 | ENST00000368325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600164380.hg.1 | ENST00000368328.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600164380.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164380.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164380.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164380.hg.1 | ENST00000368325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600164380.hg.1 | ENST00000368328.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600164380.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164380.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164380.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164380.hg.1 | TC0600009381.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 228202 | - | - |

CENPW
Genomic Map