Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009892.hg.1 |
ENST00000444158.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009892.hg.1 |
ENST00000444158.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009892.hg.1 |
ENST00000444158.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009892.hg.1 |
ENST00000444158.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009892.hg.1 |
ENST00000444158.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009892.hg.1 |
ENST00000444158.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009892.hg.1 |
ENST00000444158.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009892.hg.1 |
ENST00000444158.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009892.hg.1 |
NR_110157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009892.hg.1 |
NR_110157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009892.hg.1 |
NR_110157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009892.hg.1 |
NR_110157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009892.hg.1 |
NR_110157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009892.hg.1 |
NR_110157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009892.hg.1 |
NR_110157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009892.hg.1 |
NR_110157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009892.hg.1 |
ENST00000444158.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009892.hg.1 |
ENST00000444158.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009892.hg.1 |
ENST00000444158.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009892.hg.1 |
ENST00000444158.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009892.hg.1 |
ENST00000444158.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009892.hg.1 |
ENST00000444158.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009892.hg.1 |
ENST00000444158.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009892.hg.1 |
ENST00000444158.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009892.hg.1 |
NR_110157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009892.hg.1 |
NR_110157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009892.hg.1 |
NR_110157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009892.hg.1 |
NR_110157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009892.hg.1 |
NR_110157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009892.hg.1 |
NR_110157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009892.hg.1 |
NR_110157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009892.hg.1 |
NR_110157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700170381.hg.1 |
ENST00000444158.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700170381.hg.1 |
NR_110157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700170381.hg.1 |
ENST00000444158.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700170381.hg.1 |
NR_110157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700170381.hg.1 |
TC0700009892.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700170383.hg.1 |
ENST00000444158.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700170383.hg.1 |
NR_110157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700170383.hg.1 |
ENST00000444158.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700170383.hg.1 |
NR_110157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700170383.hg.1 |
TC0700009892.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700170382.hg.1 |
ENST00000444158.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700170382.hg.1 |
NR_110157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700170382.hg.1 |
ENST00000444158.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700170382.hg.1 |
NR_110157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700170382.hg.1 |
TC0700009892.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700170384.hg.1 |
ENST00000444158.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700170384.hg.1 |
NR_110157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700170384.hg.1 |
ENST00000444158.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700170384.hg.1 |
NR_110157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700170384.hg.1 |
TC0700009892.hg.1 |
Clariom D TC |