Assay Details
Gene Symbol: LINC00326 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC039371.1 | - | 3 - 4 | 1798 | 147 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0600009496.hg.1 | ENST00000434443.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009496.hg.1 | ENST00000434443.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009496.hg.1 | ENST00000434443.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009496.hg.1 | ENST00000434443.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009496.hg.1 | ENST00000457339.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009496.hg.1 | ENST00000457339.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009496.hg.1 | ENST00000457339.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009496.hg.1 | ENST00000457339.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009496.hg.1 | ENST00000457339.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009496.hg.1 | ENST00000457339.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009496.hg.1 | ENST00000457339.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009496.hg.1 | ENST00000457339.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009496.hg.1 | ENST00000457339.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009496.hg.1 | ENST00000457339.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009496.hg.1 | ENST00000457339.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009496.hg.1 | ENST00000457339.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009496.hg.1 | ENST00000457339.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009496.hg.1 | ENST00000457339.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009496.hg.1 | ENST00000457339.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009496.hg.1 | ENST00000457339.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009496.hg.1 | ENST00000457339.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009496.hg.1 | ENST00000457339.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009496.hg.1 | NR_026969.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009496.hg.1 | NR_026969.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009496.hg.1 | NR_026969.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009496.hg.1 | NR_026969.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009496.hg.1 | NR_026969.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009496.hg.1 | NR_026969.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009496.hg.1 | ENST00000434443.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009496.hg.1 | ENST00000434443.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009496.hg.1 | ENST00000434443.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009496.hg.1 | ENST00000434443.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009496.hg.1 | ENST00000457339.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009496.hg.1 | ENST00000457339.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009496.hg.1 | ENST00000457339.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009496.hg.1 | ENST00000457339.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009496.hg.1 | ENST00000457339.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009496.hg.1 | ENST00000457339.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009496.hg.1 | ENST00000457339.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009496.hg.1 | ENST00000457339.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009496.hg.1 | ENST00000457339.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009496.hg.1 | ENST00000457339.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009496.hg.1 | ENST00000457339.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009496.hg.1 | ENST00000457339.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009496.hg.1 | ENST00000457339.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009496.hg.1 | ENST00000457339.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009496.hg.1 | ENST00000457339.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009496.hg.1 | ENST00000457339.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009496.hg.1 | ENST00000457339.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009496.hg.1 | ENST00000457339.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009496.hg.1 | NR_026969.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009496.hg.1 | NR_026969.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009496.hg.1 | NR_026969.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009496.hg.1 | NR_026969.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009496.hg.1 | NR_026969.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009496.hg.1 | NR_026969.1 | Refseq |
