Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010948.hg.1 |
ENST00000451496.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010948.hg.1 |
ENST00000451496.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010948.hg.1 |
ENST00000451496.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010948.hg.1 |
NR_111908.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010948.hg.1 |
NR_111908.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010948.hg.1 |
NR_111908.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500176335.hg.1 |
ENST00000451496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500176335.hg.1 |
NR_111908.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500176335.hg.1 |
ENST00000451496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500176335.hg.1 |
NR_111908.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500176335.hg.1 |
TC0500010948.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500176336.hg.1 |
NR_111908.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500176336.hg.1 |
NR_111908.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500176336.hg.1 |
TC0500010948.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500010948.hg.1 |
ENST00000451496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010948.hg.1 |
ENST00000451496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010948.hg.1 |
ENST00000451496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010948.hg.1 |
NR_111908.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010948.hg.1 |
NR_111908.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010948.hg.1 |
NR_111908.1 |
Refseq |