Assay Details
| Gene Symbol: MYH16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002147.2 | 9 - 10 | 1262 | 88 | ||
| GenBank mRNA | AK025690.1 | - | 9 - 10 | 1262 | 88 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | karyopherin subunit alpha 7 | KPNA7 | IPOA8 | -68.6kb | Anti-sense | Chr. 7 - 99145467 - 99219417 on NCBI Build GRCh38 |
| Downstream | actin related protein 2/3 complex subunit 1A | ARPC1A |
Arc40, HEL-68, HEL-S-307, SOP2Hs, SOP2L |
36.5kb | Sense | Chr. 7 - 99325873 - 99366262 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07000591.hg.1
Set Membership: |
|

MYH16
Genomic Map