Assay Details
Gene Symbol: SYCE1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129979.1 | NP_001123451.1 | 4 - 5 | 249 | 62 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC16002059.hg.1, TC16002060.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR16008884.hg.1 | TC16002059.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR16008884.hg.1 | TC16002060.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR16008884.hg.1 | ENST00000248248.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16008884.hg.1 | ENST00000378644.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16008884.hg.1 | NM_001129979.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16008884.hg.1 | NM_014940.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16008888.hg.1 | TC16002060.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR16008888.hg.1 | ENST00000378644.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16008888.hg.1 | NM_001129979.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16008890.hg.1 | TC16002060.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR16008890.hg.1 | ENST00000378644.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16008890.hg.1 | NM_001129979.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16008887.hg.1 | TC16002060.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR16008887.hg.1 | ENST00000378644.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16008887.hg.1 | NM_001129979.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000563157.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000563157.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000563157.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000568925.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000568925.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000568925.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000568925.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000568925.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000568925.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16008889.hg.1 | TC16002060.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR16008889.hg.1 | ENST00000378644.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16008889.hg.1 | NM_001129979.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600191553.hg.1 | ENST00000378644.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600191553.hg.1 | NM_001129979.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600191553.hg.1 | ENST00000378644.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600191553.hg.1 | NM_001129979.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600191553.hg.1 | TC1600011433.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1600191554.hg.1 | ENST00000378644.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600191554.hg.1 | NM_001129979.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600191554.hg.1 | ENST00000378644.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600191554.hg.1 | NM_001129979.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600191554.hg.1 | TC1600011433.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1600191556.hg.1 | ENST00000378644.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600191556.hg.1 | NM_001129979.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600191556.hg.1 | ENST00000378644.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600191556.hg.1 | NM_001129979.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600191556.hg.1 | TC1600011433.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000563157.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000563157.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000563157.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000568925.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000568925.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000568925.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000568925.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000568925.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000568925.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600191552.hg.1 | ENST00000378644.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600191552.hg.1 | NM_001129979.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600191552.hg.1 | ENST00000378644.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600191552.hg.1 | NM_001129979.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600191552.hg.1 | TC1600011433.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1600191555.hg.1 | ENST00000378644.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600191555.hg.1 | NM_001129979.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600191555.hg.1 | ENST00000378644.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600191555.hg.1 | NM_001129979.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600191555.hg.1 | TC1600011433.hg.1 | Clariom D TC |
