Assay Details
| Gene Symbol: PTPRQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145026.1 | NP_001138498.1 | 34 - 35 | 5787 | 90 | |
| XM_017019273.1 | XP_016874762.1 | 38 - 39 | 6589 | 90 | ||
| XM_017019274.1 | XP_016874763.1 | 38 - 39 | 6590 | 90 | ||
| XM_017019275.1 | XP_016874764.1 | 37 - 38 | 6516 | 90 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000668.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000547485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000547485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000547485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000547485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000547485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000547881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000547881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000547881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000547881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000547881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000549355.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000549355.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000549355.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000549355.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000551624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000551624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000551624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000551624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000614701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | ENST00000616559.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012678.hg.1 | NM_001145026.1 | Refseq |

PTPRQ
Genomic Map