Assay Details
| Gene Symbol: FREM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207361.5 | NP_997244.4 | 21 - 22 | 8977 | 67 | |
| GenBank mRNA | BX538150.1 | - | 4 - 5 | 444 | 67 | |
| BX538304.1 | - | 17 - 18 | 2941 | 67 | ||
| BX640686.1 | - | 1 - 2 | 114 | 67 | ||
| CR933724.1 | - | 9 - 10 | 1469 | 67 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13000132.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR13001542.hg.1 | ENST00000280481.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13001542.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13001542.hg.1 | TC13000132.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000482551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000482551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000482551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000482551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000482551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000482551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000482551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000482551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300147025.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300147025.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300147025.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300147025.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300147025.hg.1 | TC1300006919.hg.1 | Clariom D TC |

FREM2
Genomic Map