| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12012224.hg.1 |
TC12003258.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12012224.hg.1 |
ENST00000551765.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12012224.hg.1 |
NM_176818.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12012224.hg.1 |
NR_033684.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12012227.hg.1 |
TC12003258.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12012227.hg.1 |
NR_033684.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012717.hg.1 |
ENST00000229384.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012717.hg.1 |
ENST00000229384.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012717.hg.1 |
ENST00000229384.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012717.hg.1 |
ENST00000548171.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012717.hg.1 |
ENST00000548171.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012717.hg.1 |
ENST00000548171.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012717.hg.1 |
ENST00000548171.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012717.hg.1 |
ENST00000548171.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012717.hg.1 |
ENST00000548171.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012717.hg.1 |
ENST00000548171.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012717.hg.1 |
ENST00000551765.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012717.hg.1 |
ENST00000551765.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012717.hg.1 |
ENST00000551765.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012717.hg.1 |
ENST00000551765.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012717.hg.1 |
ENST00000551765.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012717.hg.1 |
ENST00000551765.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012717.hg.1 |
ENST00000551765.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012717.hg.1 |
NM_176818.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012717.hg.1 |
NM_176818.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012717.hg.1 |
NM_176818.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012717.hg.1 |
NM_176818.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012717.hg.1 |
NM_176818.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012717.hg.1 |
NM_176818.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012717.hg.1 |
NM_176818.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012717.hg.1 |
NR_033684.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012717.hg.1 |
NR_033684.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012717.hg.1 |
NR_033684.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012717.hg.1 |
NR_033684.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012717.hg.1 |
NR_033684.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012717.hg.1 |
NR_033684.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012717.hg.1 |
NR_033684.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012717.hg.1 |
NR_033684.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12012226.hg.1 |
TC12003258.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12012226.hg.1 |
ENST00000551765.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12012226.hg.1 |
NM_176818.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12012226.hg.1 |
NR_033684.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12012225.hg.1 |
TC12003258.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12012225.hg.1 |
ENST00000551765.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12012225.hg.1 |
NM_176818.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12012225.hg.1 |
NR_033684.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200201776.hg.1 |
ENST00000229384.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200201776.hg.1 |
ENST00000548171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200201776.hg.1 |
ENST00000551765.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200201776.hg.1 |
NM_176818.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200201776.hg.1 |
NR_033684.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200201776.hg.1 |
ENST00000229384.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200201776.hg.1 |
ENST00000548171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200201776.hg.1 |
ENST00000551765.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200201776.hg.1 |
NM_176818.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200201776.hg.1 |
NR_033684.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200201776.hg.1 |
TC1200012717.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200201773.hg.1 |
ENST00000548171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200201773.hg.1 |
ENST00000551765.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200201773.hg.1 |
NM_176818.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200201773.hg.1 |
NR_033684.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200201773.hg.1 |
ENST00000548171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200201773.hg.1 |
ENST00000551765.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200201773.hg.1 |
NM_176818.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200201773.hg.1 |
NR_033684.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200201773.hg.1 |
TC1200012717.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1200012717.hg.1 |
ENST00000229384.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012717.hg.1 |
ENST00000229384.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012717.hg.1 |
ENST00000229384.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012717.hg.1 |
ENST00000548171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012717.hg.1 |
ENST00000548171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012717.hg.1 |
ENST00000548171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012717.hg.1 |
ENST00000548171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012717.hg.1 |
ENST00000548171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012717.hg.1 |
ENST00000548171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012717.hg.1 |
ENST00000548171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012717.hg.1 |
ENST00000551765.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012717.hg.1 |
ENST00000551765.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012717.hg.1 |
ENST00000551765.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012717.hg.1 |
ENST00000551765.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012717.hg.1 |
ENST00000551765.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012717.hg.1 |
ENST00000551765.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012717.hg.1 |
ENST00000551765.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012717.hg.1 |
NM_176818.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012717.hg.1 |
NM_176818.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012717.hg.1 |
NM_176818.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012717.hg.1 |
NM_176818.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012717.hg.1 |
NM_176818.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012717.hg.1 |
NM_176818.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012717.hg.1 |
NM_176818.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012717.hg.1 |
NR_033684.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012717.hg.1 |
NR_033684.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012717.hg.1 |
NR_033684.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012717.hg.1 |
NR_033684.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012717.hg.1 |
NR_033684.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012717.hg.1 |
NR_033684.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012717.hg.1 |
NR_033684.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012717.hg.1 |
NR_033684.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200201774.hg.1 |
ENST00000548171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200201774.hg.1 |
ENST00000551765.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200201774.hg.1 |
NM_176818.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200201774.hg.1 |
NR_033684.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200201774.hg.1 |
ENST00000548171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200201774.hg.1 |
ENST00000551765.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200201774.hg.1 |
NM_176818.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200201774.hg.1 |
NR_033684.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200201774.hg.1 |
TC1200012717.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200201777.hg.1 |
ENST00000548171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200201777.hg.1 |
NR_033684.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200201777.hg.1 |
ENST00000548171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200201777.hg.1 |
NR_033684.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200201777.hg.1 |
TC1200012717.hg.1 |
Clariom D TC |