Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009427.hg.1 |
ENST00000430373.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009427.hg.1 |
ENST00000430373.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009427.hg.1 |
ENST00000430373.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009427.hg.1 |
ENST00000430373.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009427.hg.1 |
ENST00000524935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009427.hg.1 |
ENST00000524935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009427.hg.1 |
NR_038951.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009427.hg.1 |
NR_038951.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009427.hg.1 |
NR_038951.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009427.hg.1 |
NR_038951.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009427.hg.1 |
NR_038952.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009427.hg.1 |
NR_038952.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009427.hg.1 |
NR_038952.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015643.hg.1 |
TC01000980.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015643.hg.1 |
ENST00000430373.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015643.hg.1 |
NR_038951.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015642.hg.1 |
TC01000980.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015642.hg.1 |
ENST00000430373.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015642.hg.1 |
NR_038951.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015642.hg.1 |
NR_038952.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015644.hg.1 |
TC01000980.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015644.hg.1 |
ENST00000430373.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015644.hg.1 |
NR_038951.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015644.hg.1 |
NR_038952.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100167840.hg.1 |
ENST00000430373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100167840.hg.1 |
ENST00000524935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100167840.hg.1 |
NR_038951.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100167840.hg.1 |
NR_038952.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100167840.hg.1 |
ENST00000430373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100167840.hg.1 |
ENST00000524935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100167840.hg.1 |
NR_038951.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100167840.hg.1 |
NR_038952.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100167840.hg.1 |
TC0100009427.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100167843.hg.1 |
ENST00000430373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100167843.hg.1 |
NR_038951.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100167843.hg.1 |
NR_038952.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100167843.hg.1 |
ENST00000430373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100167843.hg.1 |
NR_038951.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100167843.hg.1 |
NR_038952.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100167843.hg.1 |
TC0100009427.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100167841.hg.1 |
ENST00000430373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100167841.hg.1 |
NR_038951.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100167841.hg.1 |
ENST00000430373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100167841.hg.1 |
NR_038951.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100167841.hg.1 |
TC0100009427.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100009427.hg.1 |
ENST00000430373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009427.hg.1 |
ENST00000430373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009427.hg.1 |
ENST00000430373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009427.hg.1 |
ENST00000430373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009427.hg.1 |
ENST00000524935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009427.hg.1 |
ENST00000524935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009427.hg.1 |
NR_038951.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009427.hg.1 |
NR_038951.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009427.hg.1 |
NR_038951.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009427.hg.1 |
NR_038951.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009427.hg.1 |
NR_038952.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009427.hg.1 |
NR_038952.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009427.hg.1 |
NR_038952.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100167842.hg.1 |
ENST00000430373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100167842.hg.1 |
ENST00000524935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100167842.hg.1 |
NR_038951.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100167842.hg.1 |
NR_038952.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100167842.hg.1 |
ENST00000430373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100167842.hg.1 |
ENST00000524935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100167842.hg.1 |
NR_038951.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100167842.hg.1 |
NR_038952.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100167842.hg.1 |
TC0100009427.hg.1 |
Clariom D TC |