| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Clariom D Assay |
PSR1200162646.hg.1 |
ENST00000342887.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200162646.hg.1 |
ENST00000524981.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200162646.hg.1 |
NM_001306084.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200162646.hg.1 |
ENST00000342887.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200162646.hg.1 |
ENST00000524981.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200162646.hg.1 |
NM_001306084.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200162646.hg.1 |
TC1200008541.hg.1 |
Clariom D TC |