Assay Details
Gene Symbol: LOC283352 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_104172.1 | 2 - 3 | 210 | 75 | ||
GenBank mRNA | BC035922.1 | - | 1 - 2 | 51 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000510001.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000510001.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000510001.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000510001.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000510001.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000510001.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000540739.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000540739.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000540739.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000540739.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | NR_104172.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | NR_104172.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | NR_104172.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | NR_104172.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | NR_104172.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | NR_104172.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | NR_104173.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | NR_104173.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | NR_104173.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | NR_104173.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | NR_104173.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000510001.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000510001.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000510001.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000510001.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000510001.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000510001.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000540739.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000540739.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000540739.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000540739.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | NR_104172.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | NR_104172.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | NR_104172.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | NR_104172.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | NR_104172.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | NR_104172.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | NR_104173.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | NR_104173.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | NR_104173.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | NR_104173.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | NR_104173.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200169843.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200169843.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200169843.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200169843.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200169843.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200169843.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200169843.hg.1 | ENST00000540739.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200169843.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200169843.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200169843.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200169843.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200169843.hg.1 | NR_104172.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200169843.hg.1 | NR_104173.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200169843.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200169843.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200169843.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200169843.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200169843.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200169843.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200169843.hg.1 | ENST00000540739.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200169843.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200169843.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200169843.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200169843.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200169843.hg.1 | NR_104172.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200169843.hg.1 | NR_104173.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200169843.hg.1 | TC1200009378.hg.1 | Clariom D TC |
