Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013184.hg.1 |
TC0X000993.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013184.hg.1 |
ENST00000377879.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013184.hg.1 |
NR_046101.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009545.hg.1 |
ENST00000377879.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009545.hg.1 |
ENST00000377879.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009545.hg.1 |
ENST00000377879.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009545.hg.1 |
ENST00000377879.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009545.hg.1 |
ENST00000618037.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009545.hg.1 |
ENST00000618037.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009545.hg.1 |
ENST00000618037.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009545.hg.1 |
NR_046101.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009545.hg.1 |
NR_046101.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009545.hg.1 |
NR_046101.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009545.hg.1 |
NR_046101.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013183.hg.1 |
TC0X000993.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013183.hg.1 |
ENST00000377879.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013183.hg.1 |
NR_046101.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013182.hg.1 |
TC0X000993.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013182.hg.1 |
ENST00000377879.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013182.hg.1 |
NR_046101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00165139.hg.1 |
ENST00000377879.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00165139.hg.1 |
NR_046101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00165139.hg.1 |
ENST00000377879.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00165139.hg.1 |
NR_046101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00165139.hg.1 |
TC0X00009545.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00165137.hg.1 |
ENST00000377879.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00165137.hg.1 |
ENST00000618037.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00165137.hg.1 |
NR_046101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00165137.hg.1 |
ENST00000377879.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00165137.hg.1 |
ENST00000618037.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00165137.hg.1 |
NR_046101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00165137.hg.1 |
TC0X00009545.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00009545.hg.1 |
ENST00000377879.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009545.hg.1 |
ENST00000377879.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009545.hg.1 |
ENST00000377879.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009545.hg.1 |
ENST00000377879.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009545.hg.1 |
ENST00000618037.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009545.hg.1 |
ENST00000618037.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009545.hg.1 |
ENST00000618037.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009545.hg.1 |
NR_046101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009545.hg.1 |
NR_046101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009545.hg.1 |
NR_046101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009545.hg.1 |
NR_046101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00165136.hg.1 |
ENST00000377879.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00165136.hg.1 |
ENST00000618037.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00165136.hg.1 |
NR_046101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00165136.hg.1 |
ENST00000377879.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00165136.hg.1 |
ENST00000618037.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00165136.hg.1 |
NR_046101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00165136.hg.1 |
TC0X00009545.hg.1 |
Clariom D TC |