Assay Details
| Gene Symbol: LOC101927616 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110057.1 | 2 - 3 | 129 | |||
| GenBank mRNA | BC039530.1 | - | 2 - 3 | 203 | 129 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927616| Gene Symbol: LOC101927616 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110057.1 | 2 - 3 | 129 | |||
| GenBank mRNA | BC039530.1 | - | 2 - 3 | 203 | 129 | |
Set Membership: |
|