Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008233.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008233.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008233.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008233.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008233.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008233.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008233.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008233.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008233.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008233.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008233.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008233.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008233.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008233.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008233.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008233.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008233.hg.1 |
ENST00000608172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008233.hg.1 |
ENST00000608172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008233.hg.1 |
ENST00000608172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008233.hg.1 |
ENST00000608172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008233.hg.1 |
ENST00000608172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008233.hg.1 |
ENST00000608172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008233.hg.1 |
ENST00000608172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008233.hg.1 |
NR_110393.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008233.hg.1 |
NR_110393.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008233.hg.1 |
NR_110393.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008233.hg.1 |
NR_110393.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008233.hg.1 |
NR_110393.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008233.hg.1 |
NR_110393.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008233.hg.1 |
NR_110393.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00156247.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156247.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156247.hg.1 |
ENST00000608172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156247.hg.1 |
NR_110393.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00156247.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156247.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156247.hg.1 |
ENST00000608172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156247.hg.1 |
NR_110393.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00156247.hg.1 |
TC0X00008233.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00156251.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156251.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156251.hg.1 |
ENST00000608172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156251.hg.1 |
NR_110393.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00156251.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156251.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156251.hg.1 |
ENST00000608172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156251.hg.1 |
NR_110393.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00156251.hg.1 |
TC0X00008233.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00156250.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156250.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156250.hg.1 |
ENST00000608172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156250.hg.1 |
NR_110393.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00156250.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156250.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156250.hg.1 |
ENST00000608172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156250.hg.1 |
NR_110393.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00156250.hg.1 |
TC0X00008233.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00008233.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008233.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008233.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008233.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008233.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008233.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008233.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008233.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008233.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008233.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008233.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008233.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008233.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008233.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008233.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008233.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008233.hg.1 |
ENST00000608172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008233.hg.1 |
ENST00000608172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008233.hg.1 |
ENST00000608172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008233.hg.1 |
ENST00000608172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008233.hg.1 |
ENST00000608172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008233.hg.1 |
ENST00000608172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008233.hg.1 |
ENST00000608172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008233.hg.1 |
NR_110393.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008233.hg.1 |
NR_110393.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008233.hg.1 |
NR_110393.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008233.hg.1 |
NR_110393.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008233.hg.1 |
NR_110393.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008233.hg.1 |
NR_110393.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008233.hg.1 |
NR_110393.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00156249.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156249.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156249.hg.1 |
ENST00000608172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156249.hg.1 |
NR_110393.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00156249.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156249.hg.1 |
ENST00000412422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156249.hg.1 |
ENST00000608172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156249.hg.1 |
NR_110393.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00156249.hg.1 |
TC0X00008233.hg.1 |
Clariom D TC |