Assay Details
Gene Symbol: SMIM22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253790.1 | NP_001240719.1 | 4 - 5 | 732 | 63 | |
NM_001253793.1 | NP_001240722.1 | 3 - 4 | 304 | 63 | ||
GenBank mRNA | BC035868.1 | - | 4 - 5 | 732 | 63 | |
BI767985.1 | - | 3 - 4 | 304 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P229884 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P229884 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 17245 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 42091 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_18_P12373385 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P229884 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_24_P229884 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899707 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899679 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30410614 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30410594 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899703 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899708 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899704 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899690 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899702 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899681 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899697 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899694 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16846717 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899699 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899675 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899685 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30410596 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899689 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30410638 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899683 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899696 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30410592 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899706 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899682 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16846729 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899695 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899687 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899693 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899677 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30410613 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899700 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30410632 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30410603 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899680 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899677 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30410632 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899689 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899695 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899702 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30410614 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30410594 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899708 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899700 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899681 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899699 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899680 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899706 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899683 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30410613 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899697 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16846729 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899694 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30410596 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899682 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30410638 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899707 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899675 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899679 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899693 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899704 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899703 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30410592 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899696 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16846717 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30410603 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899685 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899687 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09899690 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_24_P229884 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P229884 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P170192 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 229599_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 229599_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 229599_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 229599_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 229599_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 229599_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 229599_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 229599_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 229599_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 229599_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 229599_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 229599_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 229599_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 229599_at9 | - | - |
Human Genome U95 Set | 61881_PM_at | - | - |
Human Genome U95 Set | 61881_at | - | - |
Human Genome U133 Set | 229599_at | - | - |
Human Genome U133 Set | 229599_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A007695 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A054257 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A054258 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A007695 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A054257 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A054258 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3284429 | NR_029454.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3284429 | NR_029455.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3284429 | BC022385.2 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3284429 | BC035868.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3284429 | BC048326.1 | human_transcriptome |
