Assay Details
| Gene Symbol: CYP4V2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207352.3 | NP_997235.3 | 7 - 8 | 1290 | 89 | |
| XM_005262935.3 | XP_005262992.1 | 7 - 8 | 1075 | 89 | ||
| XM_017008037.1 | XP_016863526.1 | 6 - 7 | 1048 | 89 | ||
| GenBank mRNA | AK122600.1 | - | 7 - 8 | 1025 | 89 | |
| AK126473.1 | - | 8 - 9 | 959 | 89 | ||
| AY422002.1 | - | 7 - 8 | 1290 | 89 | ||
| BC060857.1 | - | 7 - 8 | 1156 | 30333559 | 89 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04002914.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012501.hg.1 | TC04002914.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012501.hg.1 | ENST00000274118.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012501.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012501.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012508.hg.1 | TC04002914.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012508.hg.1 | ENST00000274118.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012508.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012508.hg.1 | ENST00000502665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012508.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400189415.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400189415.hg.1 | ENST00000502665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400189415.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400189415.hg.1 | ENST00000513354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400189415.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400189415.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400189415.hg.1 | ENST00000502665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400189415.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400189415.hg.1 | ENST00000513354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400189415.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400189415.hg.1 | TC0400012874.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |

CYP4V2
Genomic Map