Assay Details
Gene Symbol: LINC00613 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_103762.1 | 2 - 3 | 223 | 121 | ||
GenBank mRNA | AW237894.1 | - | 2 - 3 | 244 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
