Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011901.hg.1 |
ENST00000504365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011901.hg.1 |
ENST00000504365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011901.hg.1 |
ENST00000504365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011901.hg.1 |
ENST00000504365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011901.hg.1 |
ENST00000505952.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011901.hg.1 |
ENST00000505952.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011901.hg.1 |
ENST00000508714.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011901.hg.1 |
ENST00000508714.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011901.hg.1 |
ENST00000508714.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011901.hg.1 |
ENST00000508714.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011901.hg.1 |
ENST00000508714.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011901.hg.1 |
ENST00000508714.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011901.hg.1 |
NR_103762.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011901.hg.1 |
NR_103762.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011901.hg.1 |
NR_103762.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011901.hg.1 |
NR_103762.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011901.hg.1 |
NR_103763.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011901.hg.1 |
NR_103763.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011901.hg.1 |
NR_103763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011901.hg.1 |
ENST00000504365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011901.hg.1 |
ENST00000504365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011901.hg.1 |
ENST00000504365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011901.hg.1 |
ENST00000504365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011901.hg.1 |
ENST00000505952.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011901.hg.1 |
ENST00000505952.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011901.hg.1 |
ENST00000508714.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011901.hg.1 |
ENST00000508714.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011901.hg.1 |
ENST00000508714.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011901.hg.1 |
ENST00000508714.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011901.hg.1 |
ENST00000508714.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011901.hg.1 |
ENST00000508714.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011901.hg.1 |
NR_103762.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011901.hg.1 |
NR_103762.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011901.hg.1 |
NR_103762.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011901.hg.1 |
NR_103762.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011901.hg.1 |
NR_103763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011901.hg.1 |
NR_103763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011901.hg.1 |
NR_103763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400181684.hg.1 |
ENST00000504365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400181684.hg.1 |
ENST00000505952.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400181684.hg.1 |
ENST00000508714.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400181684.hg.1 |
ENST00000508714.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400181684.hg.1 |
NR_103762.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400181684.hg.1 |
NR_103763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400181684.hg.1 |
ENST00000504365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400181684.hg.1 |
ENST00000505952.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400181684.hg.1 |
ENST00000508714.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400181684.hg.1 |
ENST00000508714.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400181684.hg.1 |
NR_103762.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400181684.hg.1 |
NR_103763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400181684.hg.1 |
TC0400011901.hg.1 |
Clariom D TC |