Assay Details
Gene Symbol: LINC00327 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038995.1 | 1 - 2 | 675 | 110 | ||
GenBank mRNA | AL833146.1 | - | 1 - 2 | 675 | 110 | |
BX098743.1 | - | 1 - 2 | 140 | 110 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC13000047.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR13000335.hg.1 | ENST00000443778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13000335.hg.1 | NR_038995.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13000335.hg.1 | TC13000047.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1300006588.hg.1 | ENST00000443778.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006588.hg.1 | ENST00000443778.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006588.hg.1 | ENST00000443778.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006588.hg.1 | ENST00000443778.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006588.hg.1 | ENST00000443778.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006588.hg.1 | ENST00000443778.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006588.hg.1 | ENST00000443778.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006588.hg.1 | ENST00000443778.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006588.hg.1 | ENST00000443778.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006588.hg.1 | ENST00000443778.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006588.hg.1 | ENST00000575689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006588.hg.1 | ENST00000575689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006588.hg.1 | ENST00000575689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006588.hg.1 | ENST00000575689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006588.hg.1 | ENST00000576696.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006588.hg.1 | ENST00000576696.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006588.hg.1 | ENST00000576696.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006588.hg.1 | ENST00000576696.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006588.hg.1 | ENST00000576696.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006588.hg.1 | ENST00000576696.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006588.hg.1 | ENST00000576696.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006588.hg.1 | ENST00000576696.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006588.hg.1 | NR_038995.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006588.hg.1 | NR_038995.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006588.hg.1 | NR_038995.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006588.hg.1 | NR_038995.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006588.hg.1 | NR_038995.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006588.hg.1 | NR_038995.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006588.hg.1 | NR_038995.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006588.hg.1 | NR_038995.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006588.hg.1 | NR_038995.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006588.hg.1 | NR_038995.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13000333.hg.1 | NR_038995.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13000333.hg.1 | TC13000047.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR13000334.hg.1 | ENST00000443778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13000334.hg.1 | NR_038995.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13000334.hg.1 | TC13000047.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1300006588.hg.1 | ENST00000443778.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006588.hg.1 | ENST00000443778.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006588.hg.1 | ENST00000443778.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006588.hg.1 | ENST00000443778.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006588.hg.1 | ENST00000443778.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006588.hg.1 | ENST00000443778.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006588.hg.1 | ENST00000443778.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006588.hg.1 | ENST00000443778.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006588.hg.1 | ENST00000443778.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006588.hg.1 | ENST00000443778.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006588.hg.1 | ENST00000575689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006588.hg.1 | ENST00000575689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006588.hg.1 | ENST00000575689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006588.hg.1 | ENST00000575689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006588.hg.1 | ENST00000576696.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006588.hg.1 | ENST00000576696.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006588.hg.1 | ENST00000576696.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006588.hg.1 | ENST00000576696.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006588.hg.1 | ENST00000576696.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006588.hg.1 | ENST00000576696.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006588.hg.1 | ENST00000576696.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006588.hg.1 | ENST00000576696.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006588.hg.1 | NR_038995.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006588.hg.1 | NR_038995.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006588.hg.1 | NR_038995.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006588.hg.1 | NR_038995.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006588.hg.1 | NR_038995.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006588.hg.1 | NR_038995.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006588.hg.1 | NR_038995.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006588.hg.1 | NR_038995.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006588.hg.1 | NR_038995.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006588.hg.1 | NR_038995.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300144860.hg.1 | ENST00000443778.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300144860.hg.1 | NR_038995.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300144860.hg.1 | ENST00000443778.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300144860.hg.1 | NR_038995.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300144860.hg.1 | TC1300006588.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1300144858.hg.1 | ENST00000443778.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300144858.hg.1 | NR_038995.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300144858.hg.1 | ENST00000443778.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300144858.hg.1 | NR_038995.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300144858.hg.1 | TC1300006588.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1300144859.hg.1 | ENST00000443778.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300144859.hg.1 | NR_038995.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300144859.hg.1 | ENST00000443778.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300144859.hg.1 | NR_038995.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300144859.hg.1 | TC1300006588.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1300144861.hg.1 | ENST00000443778.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300144861.hg.1 | ENST00000575689.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300144861.hg.1 | ENST00000575689.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300144861.hg.1 | ENST00000576696.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300144861.hg.1 | ENST00000576696.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300144861.hg.1 | NR_038995.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300144861.hg.1 | ENST00000443778.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300144861.hg.1 | ENST00000575689.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300144861.hg.1 | ENST00000575689.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300144861.hg.1 | ENST00000576696.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300144861.hg.1 | ENST00000576696.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300144861.hg.1 | NR_038995.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300144861.hg.1 | TC1300006588.hg.1 | Clariom D TC |
