Assay Details
| Gene Symbol: FAM234B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC132772.1 | - | 1 - 2 | 158 | 40146794 | 115 |
| BC132774.1 | - | 1 - 2 | 158 | 40146796 | 115 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC132772.1, BC132774.1 transcripts.
Set Membership: |
|

FAM234B
Genomic Map