Assay Details
Gene Symbol: FAM234B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020853.1 | NP_065904.1 | 5 - 6 | 875 | 59 | |
XM_017019706.1 | XP_016875195.1 | 7 - 8 | 1186 | 59 | ||
GenBank mRNA | AB040900.1 | - | 2 - 3 | 282 | 59 | |
AK125278.1 | - | 5 - 6 | 875 | 59 | ||
AL832902.1 | - | 2 - 3 | 180 | 59 | ||
BC039267.1 | - | 5 - 6 | 864 | 5274878 | 59 | |
BC132772.1 | - | 5 - 6 | 972 | 40146794 | 59 | |
BC132774.1 | - | 5 - 6 | 972 | 40146796 | 59 | |
CR936647.1 | - | 5 - 6 | 866 | 59 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12000187.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000416494.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000416494.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000416494.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000416494.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000416494.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000416494.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000416494.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000416494.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000416494.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000416494.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000416494.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000416494.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000416494.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000416494.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000416494.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000416494.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000416494.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000416494.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000535974.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000535974.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000537625.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000537625.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000537625.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000537625.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000537625.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000537625.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000537625.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000537625.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000537625.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000537625.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000537625.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000540455.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000540455.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000540455.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000541950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000541950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000541950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000541950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000541950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000541950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000541950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000541950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000541950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000541950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | ENST00000541950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006905.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12002358.hg.1 | ENST00000197268.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12002358.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12002358.hg.1 | TC12000187.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1200148294.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200148294.hg.1 | ENST00000416494.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200148294.hg.1 | ENST00000537625.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200148294.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200148294.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200148294.hg.1 | ENST00000416494.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200148294.hg.1 | ENST00000537625.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200148294.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200148294.hg.1 | TC1200006905.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000197268.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000416494.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000416494.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000416494.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000416494.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000416494.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000416494.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000416494.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000416494.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000416494.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000416494.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000416494.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000416494.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000416494.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000416494.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000416494.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000416494.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000416494.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000416494.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000535974.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000535974.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000537625.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000537625.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000537625.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000537625.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000537625.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000537625.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000537625.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000537625.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000537625.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000537625.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000537625.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000540455.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000540455.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000540455.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000541950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000541950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000541950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000541950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000541950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000541950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000541950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000541950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000541950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000541950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | ENST00000541950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | NM_020853.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006905.hg.1 | NM_020853.1 | Refseq |
