Assay Details
| Gene Symbol: TLCD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164407.1 | NP_001157879.1 | 3 - 4 | 466 | 81 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000982.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10191044 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10191041 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10191043 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10191037 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10191046 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30739845 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30739855 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10191038 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10191042 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30739847 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10191040 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 568978 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 18492 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3345210 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P503710 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 40850 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 20578 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P03201681 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P03201683 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3345210 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3345210 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10191086 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10191075 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30739908 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16934757 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10191078 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10191085 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10191088 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10191089 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10191083 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30739888 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10191087 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10191084 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30739887 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10191091 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10191077 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30739904 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10191093 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10191094 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10191094 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10191093 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30739888 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10191078 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10191083 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10191088 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16934757 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10191087 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10191077 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30739904 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30739908 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10191075 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30739887 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10191084 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10191089 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10191086 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10191085 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10191091 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P503710 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P503710 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03201677 | - | - |
| Agilent Human 2 cDNA Microarray (G4101A) | 3440367 | - | - |
| Agilent Human 2 cDNA Microarray (G4101A) | 3440367 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P03201677 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.1 ST Array | 8011188 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1557275_PM_a_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 241359_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR17013151.hg.1 | ENST00000330676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17013151.hg.1 | NM_001164407.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17013151.hg.1 | TC17000982.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17013150.hg.1 | NM_001164407.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17013150.hg.1 | TC17000982.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009397.hg.1 | ENST00000330676.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009397.hg.1 | ENST00000330676.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009397.hg.1 | ENST00000330676.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009397.hg.1 | ENST00000330676.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009397.hg.1 | ENST00000330676.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009397.hg.1 | ENST00000330676.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009397.hg.1 | ENST00000330676.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009397.hg.1 | NM_001164407.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009397.hg.1 | NM_001164407.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009397.hg.1 | NM_001164407.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009397.hg.1 | NM_001164407.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009397.hg.1 | NM_001164407.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009397.hg.1 | NM_001164407.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700171949.hg.1 | ENST00000330676.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700171949.hg.1 | NM_001164407.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700171949.hg.1 | ENST00000330676.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700171949.hg.1 | NM_001164407.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700171949.hg.1 | TC1700009397.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700009397.hg.1 | ENST00000330676.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009397.hg.1 | ENST00000330676.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009397.hg.1 | ENST00000330676.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009397.hg.1 | ENST00000330676.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009397.hg.1 | ENST00000330676.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009397.hg.1 | ENST00000330676.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009397.hg.1 | ENST00000330676.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009397.hg.1 | NM_001164407.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009397.hg.1 | NM_001164407.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009397.hg.1 | NM_001164407.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009397.hg.1 | NM_001164407.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009397.hg.1 | NM_001164407.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009397.hg.1 | NM_001164407.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700171950.hg.1 | ENST00000330676.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700171950.hg.1 | NM_001164407.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700171950.hg.1 | ENST00000330676.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700171950.hg.1 | NM_001164407.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700171950.hg.1 | TC1700009397.hg.1 | Clariom D TC |
| Human Genome U95 Set | 83388_at | - | - |
| Human Genome U95 Set | 83388_PM_at | - | - |
| Human Genome U133 Set | 241359_at | - | - |
| Human Genome U133 Set | 241359_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A054986 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A054986 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 199902 | - | - |

TLCD2
Genomic Map