Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR18003990.hg.1 |
NR_040031.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18003990.hg.1 |
TC18000379.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008151.hg.1 |
ENST00000588672.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008151.hg.1 |
ENST00000588672.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008151.hg.1 |
ENST00000588672.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008151.hg.1 |
ENST00000588672.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008151.hg.1 |
ENST00000588672.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008151.hg.1 |
NR_040031.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008151.hg.1 |
NR_040031.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008151.hg.1 |
NR_040031.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008151.hg.1 |
NR_040031.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008151.hg.1 |
NR_040031.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18003991.hg.1 |
NR_040031.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18003991.hg.1 |
TC18000379.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1800155745.hg.1 |
ENST00000588672.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800155745.hg.1 |
NR_040031.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800155745.hg.1 |
TC1800008151.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800155745.hg.1 |
ENST00000588672.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800155745.hg.1 |
NR_040031.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800155744.hg.1 |
ENST00000588672.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800155744.hg.1 |
NR_040031.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800155744.hg.1 |
TC1800008151.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800155744.hg.1 |
ENST00000588672.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800155744.hg.1 |
NR_040031.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008151.hg.1 |
ENST00000588672.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008151.hg.1 |
ENST00000588672.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008151.hg.1 |
ENST00000588672.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008151.hg.1 |
ENST00000588672.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008151.hg.1 |
ENST00000588672.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008151.hg.1 |
NR_040031.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008151.hg.1 |
NR_040031.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008151.hg.1 |
NR_040031.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008151.hg.1 |
NR_040031.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008151.hg.1 |
NR_040031.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800155746.hg.1 |
NR_040031.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800155746.hg.1 |
TC1800008151.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800155746.hg.1 |
NR_040031.1 |
Refseq |