Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR18003988.hg.1 |
NR_040031.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18003988.hg.1 |
TC18000379.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008151.hg.1 |
ENST00000588672.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008151.hg.1 |
ENST00000588672.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008151.hg.1 |
ENST00000588672.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008151.hg.1 |
ENST00000588672.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008151.hg.1 |
ENST00000588672.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008151.hg.1 |
NR_040031.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008151.hg.1 |
NR_040031.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008151.hg.1 |
NR_040031.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008151.hg.1 |
NR_040031.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008151.hg.1 |
NR_040031.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800155742.hg.1 |
ENST00000588672.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800155742.hg.1 |
NR_040031.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800155742.hg.1 |
TC1800008151.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800155742.hg.1 |
ENST00000588672.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800155742.hg.1 |
NR_040031.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008151.hg.1 |
ENST00000588672.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008151.hg.1 |
ENST00000588672.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008151.hg.1 |
ENST00000588672.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008151.hg.1 |
ENST00000588672.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008151.hg.1 |
ENST00000588672.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008151.hg.1 |
NR_040031.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008151.hg.1 |
NR_040031.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008151.hg.1 |
NR_040031.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008151.hg.1 |
NR_040031.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008151.hg.1 |
NR_040031.1 |
Refseq |
Human Genome U133 Plus 2.0 Array |
1560181_PM_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
1560181_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1560181_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1560181_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1560181_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1560181_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1560181_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1560181_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1560181_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1560181_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1560181_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1560181_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1560181_at4 |
- |
- |