Assay Details
| Gene Symbol: ZAR1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136571.1 | NP_001130043.1 | 1 - 2 | 683 | 124 | |
| GenBank mRNA | BC146956.1 | - | 1 - 2 | 681 | 9056884 | 124 |
| BC146965.1 | - | 1 - 2 | 683 | 9056893 | 124 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13000548.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 53275 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 120315 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 639666 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3313476 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_1897 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_1898 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 3995 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_18_P11690535 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_18_P11690538 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_18_P11684406 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P02750122 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3313476 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3313476 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08761577 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29208384 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29208374 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08761592 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08761585 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29208386 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29208391 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08761596 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16624054 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08761575 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08761588 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29208383 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08761578 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29208385 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29208371 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08761590 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08761583 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08761595 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16624058 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29208372 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16624060 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29208372 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08761588 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08761578 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08761596 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16624060 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08761595 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29208385 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16624054 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29208371 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08761590 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29208383 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08761575 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08761577 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29208384 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08761585 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16624058 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29208391 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08761592 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29208374 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08761583 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29208386 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16624052 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16624069 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08761589 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08761617 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08761625 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08761569 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08761590 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08761596 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08761579 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08761615 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08761584 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08761602 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08761564 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08761607 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08761592 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08761573 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08761634 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08761627 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08761632 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08761611 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08761566 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08761619 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08761567 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08761636 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08761574 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08761610 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08761578 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08761604 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08761599 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16624059 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08761623 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08761559 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16624062 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08761555 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16624063 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08761630 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16624055 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08761633 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08761622 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08761553 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16624053 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08761561 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08761571 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08761613 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16624060 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P02750122 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P19767058 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 7970889 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7970891 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7970890 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7970888 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7970887 | - | - |
| Affymetrix Human Genome U219 Array | 11735569_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11735569_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR13006144.hg.1 | ENST00000345108.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13006144.hg.1 | NM_001136571.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13006144.hg.1 | TC13000548.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR13006142.hg.1 | ENST00000345108.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13006142.hg.1 | NM_001136571.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13006142.hg.1 | TC13000548.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR13006143.hg.1 | ENST00000345108.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13006143.hg.1 | NM_001136571.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13006143.hg.1 | TC13000548.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR13006141.hg.1 | ENST00000345108.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13006141.hg.1 | NM_001136571.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13006141.hg.1 | TC13000548.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008552.hg.1 | ENST00000345108.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008552.hg.1 | ENST00000345108.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008552.hg.1 | ENST00000345108.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008552.hg.1 | ENST00000345108.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008552.hg.1 | ENST00000345108.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008552.hg.1 | ENST00000345108.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008552.hg.1 | ENST00000533490.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008552.hg.1 | ENST00000533490.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008552.hg.1 | ENST00000533490.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008552.hg.1 | ENST00000533490.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008552.hg.1 | ENST00000533490.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008552.hg.1 | ENST00000533490.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008552.hg.1 | ENST00000533490.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008552.hg.1 | ENST00000533490.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008552.hg.1 | ENST00000533490.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008552.hg.1 | ENST00000533490.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008552.hg.1 | NM_001136571.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008552.hg.1 | NM_001136571.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008552.hg.1 | NM_001136571.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008552.hg.1 | NM_001136571.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008552.hg.1 | NM_001136571.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008552.hg.1 | NM_001136571.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300157633.hg.1 | ENST00000345108.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157633.hg.1 | ENST00000533490.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157633.hg.1 | NM_001136571.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300157633.hg.1 | ENST00000345108.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157633.hg.1 | ENST00000533490.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157633.hg.1 | NM_001136571.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300157633.hg.1 | TC1300008552.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300157634.hg.1 | ENST00000345108.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157634.hg.1 | ENST00000533490.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157634.hg.1 | NM_001136571.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300157634.hg.1 | ENST00000345108.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157634.hg.1 | ENST00000533490.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157634.hg.1 | NM_001136571.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300157634.hg.1 | TC1300008552.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300157635.hg.1 | ENST00000345108.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157635.hg.1 | ENST00000533490.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157635.hg.1 | NM_001136571.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300157635.hg.1 | ENST00000345108.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157635.hg.1 | ENST00000533490.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157635.hg.1 | NM_001136571.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300157635.hg.1 | TC1300008552.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300157632.hg.1 | ENST00000345108.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157632.hg.1 | ENST00000533490.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157632.hg.1 | NM_001136571.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300157632.hg.1 | ENST00000345108.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157632.hg.1 | ENST00000533490.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157632.hg.1 | NM_001136571.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300157632.hg.1 | TC1300008552.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1300008552.hg.1 | ENST00000345108.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008552.hg.1 | ENST00000345108.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008552.hg.1 | ENST00000345108.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008552.hg.1 | ENST00000345108.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008552.hg.1 | ENST00000345108.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008552.hg.1 | ENST00000345108.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008552.hg.1 | ENST00000533490.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008552.hg.1 | ENST00000533490.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008552.hg.1 | ENST00000533490.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008552.hg.1 | ENST00000533490.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008552.hg.1 | ENST00000533490.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008552.hg.1 | ENST00000533490.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008552.hg.1 | ENST00000533490.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008552.hg.1 | ENST00000533490.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008552.hg.1 | ENST00000533490.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008552.hg.1 | ENST00000533490.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008552.hg.1 | NM_001136571.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008552.hg.1 | NM_001136571.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008552.hg.1 | NM_001136571.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008552.hg.1 | NM_001136571.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008552.hg.1 | NM_001136571.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008552.hg.1 | NM_001136571.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300157636.hg.1 | ENST00000345108.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157636.hg.1 | ENST00000533490.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157636.hg.1 | NM_001136571.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300157636.hg.1 | ENST00000345108.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157636.hg.1 | ENST00000533490.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157636.hg.1 | NM_001136571.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300157636.hg.1 | TC1300008552.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300157637.hg.1 | ENST00000345108.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157637.hg.1 | ENST00000533490.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157637.hg.1 | NM_001136571.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300157637.hg.1 | ENST00000345108.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157637.hg.1 | ENST00000533490.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157637.hg.1 | NM_001136571.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300157637.hg.1 | TC1300008552.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A049404 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A049404 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 219104 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1668436 | NM_001136571.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1668436 | BC146956.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1668436 | BC146965.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1668436 | NM_001136571.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1668436 | BC146956.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1668436 | BC146965.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1668436 | NM_001136571.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1668436 | BC146956.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1668436 | BC146965.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1668436 | NM_001136571.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1668436 | BC146956.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1668436 | BC146965.1 | human_transcriptome |

ZAR1L
Genomic Map