Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008886.hg.1 |
ENST00000433480.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008886.hg.1 |
ENST00000433480.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008886.hg.1 |
ENST00000433480.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008886.hg.1 |
ENST00000433480.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008886.hg.1 |
ENST00000433480.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008886.hg.1 |
ENST00000433480.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008886.hg.1 |
ENST00000433480.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008886.hg.1 |
ENST00000436963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008886.hg.1 |
ENST00000436963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008886.hg.1 |
NR_046414.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008886.hg.1 |
NR_046414.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008886.hg.1 |
NR_046414.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008886.hg.1 |
NR_046414.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008886.hg.1 |
NR_046414.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008886.hg.1 |
NR_046414.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008886.hg.1 |
NR_046414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300159984.hg.1 |
ENST00000433480.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300159984.hg.1 |
NR_046414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300159984.hg.1 |
ENST00000433480.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300159984.hg.1 |
NR_046414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300159984.hg.1 |
TC1300008886.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300159985.hg.1 |
ENST00000433480.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300159985.hg.1 |
NR_046414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300159985.hg.1 |
ENST00000433480.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300159985.hg.1 |
NR_046414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300159985.hg.1 |
TC1300008886.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300008886.hg.1 |
ENST00000433480.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008886.hg.1 |
ENST00000433480.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008886.hg.1 |
ENST00000433480.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008886.hg.1 |
ENST00000433480.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008886.hg.1 |
ENST00000433480.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008886.hg.1 |
ENST00000433480.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008886.hg.1 |
ENST00000433480.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008886.hg.1 |
ENST00000436963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008886.hg.1 |
ENST00000436963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008886.hg.1 |
NR_046414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008886.hg.1 |
NR_046414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008886.hg.1 |
NR_046414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008886.hg.1 |
NR_046414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008886.hg.1 |
NR_046414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008886.hg.1 |
NR_046414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008886.hg.1 |
NR_046414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300159983.hg.1 |
ENST00000433480.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300159983.hg.1 |
NR_046414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300159983.hg.1 |
ENST00000433480.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300159983.hg.1 |
NR_046414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300159983.hg.1 |
TC1300008886.hg.1 |
Clariom D TC |