Assay Details
| Gene Symbol: LOC388456 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC039671.1 | - | 1 - 2 | 59 | 59 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | adenylate cyclase activating polypeptide 1 | ADCYAP1 | PACAP | -13.9kb | Anti-sense | Chr. 18 - 904342 - 912172 on NCBI Build GRCh38 |
| Downstream | long intergenic non-protein coding RNA 470 | LINC00470 | C18orf2 | 340.3kb | Sense | Chr. 18 - 1268311 - 1359629 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31093084 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10502814 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10502785 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10502772 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10502819 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31093158 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10502832 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31093136 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10502839 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31093109 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10502768 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10502798 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10502813 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10502780 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10502797 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10502796 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10502826 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10502782 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10502843 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10502783 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10502777 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10502803 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10502773 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31093087 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31093163 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10502810 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31093093 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31093111 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31093141 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31093097 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31093123 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31093146 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31093164 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31093166 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10502801 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10502808 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10502763 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31093165 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10502781 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31093149 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31093153 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10502769 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10502827 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31093085 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10502812 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10502771 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10502802 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10502829 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31093121 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10502825 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10502835 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31093082 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31093156 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10502779 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31093155 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31093132 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10502844 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31093140 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10502795 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10502811 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31093113 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31093133 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31093117 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10502841 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31093106 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31093110 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 683359 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3806558 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 7167 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3806558 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502819 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093123 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093112 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502835 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093122 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093093 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093139 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502772 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502773 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502801 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502809 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502802 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502780 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502832 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093166 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502785 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093164 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093102 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502795 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093106 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093153 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17031475 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093097 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093167 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502779 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502788 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093136 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502796 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093165 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502784 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502766 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502814 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502771 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093140 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502841 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093095 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093149 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093111 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093096 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502797 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502845 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093168 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502803 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502782 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502842 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093138 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093132 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093155 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502768 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093082 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093121 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502777 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093083 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093156 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502839 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502830 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502781 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093163 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093142 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502789 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502769 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502813 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502783 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093141 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17031472 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093109 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17031470 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502829 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502827 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502826 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093085 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502763 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502807 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502806 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093146 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093135 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093117 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502786 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502812 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093084 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502808 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093110 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093087 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502810 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502844 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502811 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502825 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502843 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093158 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502798 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502833 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093113 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093133 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502768 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502813 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502783 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093158 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502832 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502789 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502825 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502835 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502839 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093138 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093146 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093165 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093133 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093102 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502771 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502833 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093153 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502814 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502819 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093135 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502773 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093163 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502811 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093167 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093096 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17031470 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502808 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093164 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502797 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502788 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093121 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502807 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502786 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093149 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093140 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502801 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502772 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093095 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502796 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502842 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502781 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502829 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502777 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093083 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502782 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093155 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093142 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093166 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093085 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093113 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502827 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502798 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093156 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093109 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502785 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17031475 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093111 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502784 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502841 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502763 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502844 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502845 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093087 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502766 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502812 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502809 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502795 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502769 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502802 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093082 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502810 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093112 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502780 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502843 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093110 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502803 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17031472 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093123 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502826 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093136 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502779 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502806 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093122 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093093 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093132 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10502830 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093084 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093106 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093117 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093141 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093139 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093168 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31093097 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03303160 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 1563117_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007874.hg.1 | ENST00000564872.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007874.hg.1 | ENST00000564872.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007874.hg.1 | ENST00000564872.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007874.hg.1 | ENST00000564872.1 | Ensembl |
| Human Genome U133 Plus 2.0 Array | 1563117_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1563117_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563117_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563117_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563117_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563117_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563117_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563117_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563117_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563117_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563117_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563117_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563117_at11 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 256999 | - | - |

LOC388456
Genomic Map